1 - 10 of 10 products displayed
  • Canine Heartworm Antibody (Monoclonal, H21G)
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA17012
  • Size 1 mL
Applications
ELISA (1:20), Immunofluorescence (1:10), Western Blot (1:10)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17012 349.00
Check your price
  • Canine Heartworm Antibody
  • Host Goat
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # PA17207
  • Size 1 mL
Applications
ELISA (1:6000)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17207 275.00
Check your price
  • Canine Heartworm Antibody (Monoclonal, A60Q)
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518177
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518177 289.00
Check your price
  • Canine Heartworm Antibody (Monoclonal, H60H)
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518178
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518178 289.00
Check your price
  • Canine Heartworm Antibody (Monoclonal, H89A)
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518179
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518179 289.00
Check your price
  • Canine Heartworm Antibody (Monoclonal, 3221)
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA190370
  • Size 1 mL
Applications
ELISA (1:20), Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:10)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190370 550.00
Check your price
  • Canine Heartworm Antibody, HRP
  • Host Goat
  • Target Species Nematode
  • Conjugate HRP
  • Catalog # PA128353
  • Size 100 µL
Applications
ELISA (1:200-1:1000), Immunohistochemistry (1:20-1:200), Western Blot (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128353 309.00
Check your price
  • Canine Heartworm Antibody, Biotin
  • Host Goat
  • Target Species Nematode
  • Conjugate Biotin
  • Catalog # PA173112
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173112 309.00
Check your price
  • Canine Heartworm Antibody, FITC
  • Host Goat
  • Target Species Nematode
  • Conjugate FITC
  • Catalog # PA173110
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173110 309.00
Check your price
  • Canine Heartworm Antibody, HRP
  • Host Goat
  • Target Species Nematode
  • Conjugate HRP
  • Catalog # PA173111
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173111 309.00
Check your price