1 - 8 of 8 products displayed
  • Canine Parvovirus Antibody (Monoclonal, 8H7)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122686
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay-dependent)
Alternate names:
CPV, parvo
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122686 309.00
Check your price
  • Canine Parvovirus Antibody (Monoclonal, 5G7)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122685
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay-dependent)
Alternate names:
CPV, parvo
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122685 309.00
Check your price
  • Canine Parvovirus Antibody (Monoclonal, H69D)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17014
  • Size 1 mL
Applications
ELISA (1:20)
Alternate names:
CPV, parvo
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17014 349.00
Check your price
  • Canine Parvovirus Antibody (Monoclonal, A92C)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17467
  • Size 100 µg
Applications
ELISA (1:20)
Alternate names:
CPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17467 309.00
Check your price
  • Coronavirus pan Antibody (Monoclonal, FIPV3-70)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182189
  • Size 250 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
canine coronavirus, CCV, pig coronavirus, swine coronavirus, transmissible gastroenteritis virus, TGEV, ferret coronavirus, bovine coronavirus, BCV, coronavirus nucleocapsid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182189 499.00
Check your price
  • Canine/Feline Parvovirus Antibody (Monoclonal, CPVI-2AI)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182094
  • Size 250 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
CPV, parvo, FPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182094 399.00
Check your price
  • Canine/Feline Parvovirus Antibody (Monoclonal, CPV1-2A1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA191114
  • Size 250 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
CPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191114 499.00
Check your price
  • canine/Feline Parvovirus Antibody (Monoclonal, PAb416)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190661
  • Size 500 µL
Applications
Immunocytochemistry (1-5 ug/ml), Immunofluorescence (1-5 µg/ml), Immunoprecipitation (1-5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Simian vacuolating virus 40, Simian virus 40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190661 399.00
Check your price