1 - 27 of 27 products displayed
  • CEACAM1/CEACAM5 Antibody (Monoclonal, 4/3/17)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117767
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1-2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Biliary glycoprotein 1; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117767 550.00
Check your price
  • CEA Antibody (Monoclonal, CB30)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119435
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119435 309.00
Check your price
  • CEA Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128323
  • Size 50 µg
Applications
Western Blot (Assay dependent)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128323 425.00
Check your price
  • CEA Antibody (Monoclonal, 26/5/1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117766
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2µg/10^6 cells), Immunohistochemistry (Frozen) (1-2µg/10^6 cells), Western Blot (4µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117766 550.00
Check your price
  • CEACAM Pan Antibody (Monoclonal, D14HD11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117761
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunocytochemistry (1 ug/10^6 cells), Immunofluorescence (1 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Biliary glycoprotein 1; Normal cross-reacting antigen; Non-specific crossreacting antigen; Non-specific cross-reacting antigen W236; Carcinoembryonic antigen CGM7; Meconium antigen 100; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117761 550.00
Check your price
  • CEACAM5/CEACAM6 Antibody (Monoclonal, 198)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516453
  • Size 200ug
Applications
Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516453 349.00
Check your price
  • CEA Antibody, Biotin (Monoclonal, COL-1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA513711
  • Size 500 µL
Applications
Flow Cytometry (1-2 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen; Pregnancy-specific glycoprotein 30
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513711 309.00
Check your price
  • CEA Antibody (Monoclonal, COL-1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513714
  • Size 500 µL
Applications
Flow Cytometry (1-2 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen; Pregnancy-specific glycoprotein 30
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513714 249.00
Check your price
  • CEA Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516665
  • Size 500 µL
Applications
Flow Cytometry (1:10), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:500-1:1000), Immunoprecipitation (10 µg/mg lysate), Western Blot (1:200)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen; Pregnancy-specific glycoprotein 30
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516665 159.00
Check your price
  • CEA Antibody (Monoclonal, SPM506)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137419
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137419 425.00
Check your price
  • CEA Antibody (Monoclonal, 3G12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515799
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515799 349.00
Check your price
  • CEA Antibody (Monoclonal, H.426.3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515070
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515070 275.00
Check your price
  • CEA Antibody (Monoclonal, 1C7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515800
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515800 349.00
Check your price
  • CEA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532342
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532342 249.00
Check your price
  • CEA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137416
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137416 309.00
Check your price
  • CEA Antibody, FITC (Monoclonal, CI-P83-1)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA14809
  • Size 500µL
Applications
Flow Cytometry (1:25 to 1:50), Immunofluorescence (1:25 to 1:50), Immunohistochemistry (Paraffin, Frozen) (1:25 to 1:50)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA14809 399.00
Check your price
  • CEACAM Pan Antibody (Monoclonal, CLB-gran/10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110271
  • Size 100 ug
Applications
Flow Cytometry (2-8 ug/ml), Immunocytochemistry (20 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Biliary glycoprotein 1; Normal cross-reacting antigen; Non-specific crossreacting antigen; Meconium antigen 100; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110271 249.00
Check your price
  • CEA Antibody (Monoclonal, 1105)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514674
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (5-20µg/ml), Immunocytochemistry (5-20µg/ml), Immunofluorescence (5-20µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514674 349.00
Check your price
  • CEA Antibody (Monoclonal, 1106)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514675
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (5-20µg/ml), Immunocytochemistry (5-20µg/ml), Immunofluorescence (5-20µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514675 349.00
Check your price
  • CEA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513438
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513438 349.00
Check your price
  • CEA Antibody (Monoclonal, 1106)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIC0102
  • Size 1 mg
Applications
ELISA (Assay Dependent), Flow Cytometry (5-20µg/ml), Immunocytochemistry (5-20µg/ml), Immunofluorescence (5-20µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIC0102 600.00
Check your price
  • CEA Antibody (Monoclonal, 1105)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIC0101
  • Size 1 mg
Applications
ELISA (Assay Dependent), Flow Cytometry (5-20µg/ml), Immunocytochemistry (5-20µg/ml), Immunofluorescence (5-20µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIC0101 600.00
Check your price
  • CEACAM5/CEACAM6 Antibody (Monoclonal, MUS)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117762
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1-2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117762 550.00
Check your price
  • CEA Antibody (Monoclonal, 26/3/13)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117759
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1.2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117759 550.00
Check your price
  • CEA Antibody (Monoclonal, C6G9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125068
  • Size 200 µL
Applications
Immunohistochemistry (Paraffin) (1:8000), Western Blot (1:8000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125068 375.00
Check your price
  • CEA Antibody, FITC (Monoclonal, C365D3(NCRC23))
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA180578
  • Size 100 µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180578 375.00
Check your price
  • CEACAM Pan Antibody (Monoclonal, TET2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117760
  • Size 100 µg
Applications
ELISA (1:200), Flow Cytometry (1 ug/10^6 cells), Immunohistochemistry (Frozen) (1 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Non-specific cross-reacting antigen NCA-95; Carcinoembryonic antigen CGM6; CD67 antigen; Biliary glycoprotein 1; Normal cross-reacting antigen; Non-specific crossreacting antigen; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117760 550.00
Check your price