1 - 30 of 30 products displayed
  • MHC I polymorphic Antibody (Monoclonal, ER-MP42)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181746
  • Size 250 µg
Applications
Flow Cytometry (1:100-1:200), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
MHC Class 1, MHC class I polymorphic, MHC I H-2Dd/Kv/k/q/s
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181746 375.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511723
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:10-1:200)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511723 299.00
Check your price
  • MHC I Polymorphic Antibody (Monoclonal, ER-HR52)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181644
  • Size 250 µg
Applications
Flow Cytometry (1/50-1/200), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
MHC Class 1, MHC class I polymorphic, MHC I H-2b/d/p/q/w16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181644 375.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA170111
  • Size 500 ug
Applications
Flow Cytometry (1 ug/1 million cells), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170111 309.00
Check your price
  • HLA-ABC Antibody (Monoclonal, YTH862.2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180014
  • Size 200 µg
Applications
Flow Cytometry (1/50-1/100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180014 399.00
Check your price
  • MHC Class II Antibody (Monoclonal, 3D6)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA122746
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122746 375.00
Check your price
  • HLA-ABC Antibody, Biotin (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate Biotin
  • Catalog # MA180453
  • Size 100 µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180453 349.00
Check your price
  • HLA-ABC Antibody, FITC (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate FITC
  • Catalog # MA180454
  • Size 100 µg
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180454 309.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, MEM-123)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate PE
  • Catalog # MA110346
  • Size 100 ug
Applications
Flow Cytometry (1-2 ug/ml)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110346 349.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate PE
  • Catalog # MA180455
  • Size 100 Tests
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180455 349.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate PE
  • Catalog # MA119662
  • Size 100 µg
Applications
Flow Cytometry (1:50)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119662 375.00
Check your price
  • HLA-ABC Antibody (Monoclonal, YTH 862.2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190468
  • Size 100 µg
Applications
Flow Cytometry (1:50-1:100 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190468 475.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-123)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119312
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119312 199.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-147)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119151
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119151 199.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-81)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119153
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119153 199.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Rodent
  • Conjugate Unconjugated
  • Catalog # MA180123
  • Size 500 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180123 550.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119027
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119027 349.00
Check your price
  • HLA-ABC Antibody (Monoclonal, ICO-53)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17660
  • Size 100 ug
Applications
Flow Cytometry (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17660 275.00
Check your price
  • HLA-DR/DP/DQ Antibody (Monoclonal, ER-TR3)
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA140077
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140077 375.00
Check your price
  • HLA-DR/DP/DQ Antibody (Monoclonal, WR18)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180678
  • Size 200 µg
Applications
Flow Cytometry (1/50-1/200), Functional Assay (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180678 399.00
Check your price
  • MHC Class II (I-A / I-E) Antibody Monoclonal, Pacific Blue™ conjugate (M5/114.15.2) (A14901)
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # A14901
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14901 103.00
Check your price
  • MHC Class II (I-A / I-E) Antibody Monoclonal, Cy® 5.5, PerCP conjugate (M5/114.15.2) (A14902)
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # A14902
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14902 125.00
Check your price
  • HLA Class I Antibody, FITC (Monoclonal, 2G5)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Sheep, Pig, Rat
  • Conjugate FITC
  • Catalog # MA181300
  • Size 100 µg
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181300 349.00
Check your price
  • HLA Class I Antibody (Monoclonal, 2G5)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA180907
  • Size 250 µg
Applications
Flow Cytometry (1/10-1/25), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180907 425.00
Check your price
  • MHC II HLA-DR Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522113
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
MHC class II-associated invariant chain; Ia antigen-associated invariant chain; p33; HLA-DR antigens-associated invariant chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522113 349.00
Check your price
  • CD3/HLA Class 2 Antibody, FITC, PE (Monoclonal, UCHT1/WR18)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA516958
  • Size 50 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; MHC class II antigen DRB1*16; T-cell surface antigen T3/Leu-4 epsilon chain; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516958 399.00
Check your price
  • MHC II HLA-DR Antibody (Monoclonal, SPM523)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139434
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
p33; Ia antigen-associated invariant chain; HLA-DR antigens-associated invariant chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139434 425.00
Check your price
  • MHC II HLA-DR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529395
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p33; Ia antigen-associated invariant chain; HLA-DR antigens-associated invariant chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529395 349.00
Check your price
  • I-A/I-E Antibody, FITC (Monoclonal, M5/114)
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA110403
  • Size 100 ug
Applications
Flow Cytometry (4 ug/ml)
Alternate names:
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110403 225.00
Check your price
  • HLA Class I Antibody, FITC (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # AHU0228
  • Size 100 tests
Applications
Flow Cytometry (1:300)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHU0228 446.00
Check your price