1 - 6 of 6 products displayed
  • Cytomegalovirus gB Late Antigen Antibody (Monoclonal, C58G)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17292
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
CMV, HHV5gp061
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17292 309.00
Check your price
  • Cytomegalovirus gB Late Antigen Antibody (Monoclonal, I85E)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17293
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
CMV, HHV5gp061
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17293 309.00
Check your price
  • Cytomegalovirus gB Late Antigen Antibody (Monoclonal, HCMV37)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516700
  • Size 200 ug
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516700 399.00
Check your price
  • Cytomegalovirus gB Late Antigen Antibody, FITC (Monoclonal, HCMV37)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA516701
  • Size 100 ug
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516701 399.00
Check your price
  • Cytomegalovirus gB Late Antigen Antibody (Monoclonal, I55H)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17025
  • Size 1 mL
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17025 309.00
Check your price
  • Cytomegalovirus gB Late Antigen Antibody (Monoclonal, 1M12)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17598
  • Size 100 ug
Applications
ELISA (1:1000), Immunofluorescence (Assay Dependent), Western Blot (1:1000)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17598 309.00
Check your price