1 - 3 of 3 products displayed
  • Cre recombinase Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA519684
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519684 349.00
Check your price
  • Cre recombinase Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532245
  • Size 100 µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:5000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532245 349.00
Check your price
  • Cre recombinase Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532244
  • Size 100 µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532244 349.00
Check your price