1 - 8 of 8 products displayed
  • CYBA Antibody (Monoclonal, 44.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518049
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p22-phox; p22 phagocyte B-cytochrome; Superoxide-generating NADPH oxidase light chain subunit; Neutrophil cytochrome b 22 kDa polypeptide; Cytochrome b558 subunit alpha; Cytochrome b(558) alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518049 425.00
Check your price
  • CYBA Antibody (Monoclonal, CS9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518047
  • Size 100 µL
Applications
ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:1000)
Alternate names:
p22-phox; p22 phagocyte B-cytochrome; Superoxide-generating NADPH oxidase light chain subunit; Neutrophil cytochrome b 22 kDa polypeptide; Cytochrome b558 subunit alpha; Cytochrome b(558) alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518047 399.00
Check your price
  • NOX2 Antibody (Monoclonal, 54.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518052
  • Size 100 µL
Applications
Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (Assay-dependent)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518052 399.00
Check your price
  • NOX2 Antibody (Monoclonal, CL5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518050
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518050 399.00
Check your price
  • NOX2 Antibody (Monoclonal, NL7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518051
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518051 399.00
Check your price
  • NOX2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519010
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.03-0.1 µg/ml)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519010 349.00
Check your price
  • NOX2 Antibody
  • Host Rabbit
  • Target Species Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA534600
  • Size 100 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:2000), Western Blot (1 µg/ml)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534600 349.00
Check your price
  • ALDH1A2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530288
  • Size 100 µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Retinaldehyde-specific dehydrogenase type 2; Aldehyde dehydrogenase family 1 member A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530288 349.00
Check your price