1 - 30 of 30 products displayed
  • MTCO2 Antibody (Monoclonal, 12C4F12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A6404
  • Size 100 µg
Applications
Immunocytochemistry (5-10µg/ml), Western Blot (1µg/ml)
Alternate names:
Cytochrome c oxidase polypeptide II
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6404 327.00
Check your price
  • COX4 Antibody (Monoclonal, GT6310)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA517279
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517279 349.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA146154
  • Size 100 µL
Applications
Immunocytochemistry (1:40), Immunofluorescence (1:40), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:2000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146154 309.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529992
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529992 349.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521359
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521359 349.00
Check your price
  • COX4 Antibody (Monoclonal, 1D6E1A8)
  • Host Mouse
  • Target Species Bovine, C. elegans, Goat, Hamster, Human, Mollusc, Mouse, Sheep, Rhesus monkey, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # 459600
  • Size 100 µg
Applications
Western Blot (1.0 ug/ml)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
459600 359.00
Check your price
  • Complex IV Antibody (Monoclonal, 4B12A5)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # 459150
  • Size 100 µg
Applications
Western Blot (2 ug/ml)
Alternate names:
COX4, COX4-1, COXIV, COX IV-1, cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
459150 324.00
Check your price
  • COX4 Antibody (Monoclonal, K.473.4)
  • Host Rabbit
  • Target Species Bovine, Human, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515078
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:125), Immunofluorescence (1:125), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:2000), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515078 275.00
Check your price
  • COX1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526688
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Prostaglandin-endoperoxide synthase 1; Prostaglandin H2 synthase 1; Cyclooxygenase-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526688 349.00
Check your price
  • COX4 Antibody (Monoclonal, 6B3)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515686
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515686 349.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Plant, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA519471
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519471 349.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517511
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:2400), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517511 275.00
Check your price
  • COX4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518760
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV; Cytochrome c oxidase subunit IV isoform 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518760 349.00
Check your price
  • COX4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526160
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526160 349.00
Check your price
  • COX4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518759
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV; Cytochrome c oxidase subunit IV isoform 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518759 349.00
Check your price
  • COX7B2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522140
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase polypeptide VIIb2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522140 349.00
Check your price
  • COX5A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527432
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Cytochrome c oxidase polypeptide Va
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527432 349.00
Check your price
  • COX5B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521358
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase polypeptide Vb
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521358 349.00
Check your price
  • COX15 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534681
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CEMCOX2, COX15 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534681 349.00
Check your price
  • Phospho-p47phox (Ser328) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538777
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), peptide-ELISA (1:1000)
Alternate names:
p47-phox; SH3 and PX domain-containing protein 1A; Nox-organizing protein 2; Nox organizer 2; Neutrophil NADPH oxidase factor 1; NCF-47K; 47 kDa neutrophil oxidase factor; 47 kDa autosomal chronic granulomatous disease protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538777 309.00
Check your price
  • Complex IV Antibody (Monoclonal, 10G8D12C12)
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # A21347
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Western Blot (1.0 ug/ml)
Alternate names:
COX4, COX4-1, COXIV, COX IV-1, cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21347 330.00
Check your price
  • Complex IV Antibody (Monoclonal, 6E9B12D5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # 459120
  • Size 100 µg
Applications
Immunocytochemistry (5 ug/ml), Western Blot (1.0 ug/ml)
Alternate names:
COX4, COX4-1, COXIV, COX IV-1, cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
459120 330.00
Check your price
  • COX6A2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521922
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase subunit VIA-muscle; Cytochrome c oxidase polypeptide VIa-heart
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521922 349.00
Check your price
  • COX6A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526162
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit VIA-liver; Cytochrome c oxidase polypeptide VIa-liver
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526162 349.00
Check your price
  • COX5A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526161
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase polypeptide Va
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526161 349.00
Check your price
  • COX6B1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521920
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome c oxidase subunit VIb isoform 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521920 349.00
Check your price
  • NOX2 Antibody (Monoclonal, 54.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518052
  • Size 100 µL
Applications
Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (Assay-dependent)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518052 399.00
Check your price
  • COX6A2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526622
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase polypeptide VIa-heart; Cytochrome c oxidase subunit VIA-muscle
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526622 349.00
Check your price
  • Phospho-p47phox (Ser370) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536863
  • Size 100 µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p47-phox; SH3 and PX domain-containing protein 1A; Nox-organizing protein 2; Nox organizer 2; Neutrophil NADPH oxidase factor 1; NCF-47K; 47 kDa neutrophil oxidase factor; 47 kDa autosomal chronic granulomatous disease protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536863 349.00
Check your price
  • COX15 Antibody (Monoclonal, 2D2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518633
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
CEMCOX2, COX15 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518633 309.00
Check your price