1 - 2 of 2 products displayed
  • CLK3 Antibody (Monoclonal, 1H2)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA518612
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunohistochemistry (PFA fixed) (3 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
CDC-like kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518612 309.00
Check your price
  • CLK3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517289
  • Size 100 µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
CDC-like kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517289 275.00
Check your price