1 - 2 of 2 products displayed
  • Fentanyl Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175146
  • Size 100 µL
Applications
ELISA (2.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175146 600.00
Check your price
  • Phospho-OPRM1 (Ser375) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540291
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Mu opioid receptor; Mu opiate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540291 309.00
Check your price