1 - 4 of 4 products displayed
  • OXER1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132732
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor TG1019; G-protein coupled receptor R527; G-protein coupled receptor 170; 5-oxo-ETE G-protein coupled receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132732 399.00
Check your price
  • OXER1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532959
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (11 µg/ml)
Alternate names:
G-protein coupled receptor TG1019; G-protein coupled receptor R527; G-protein coupled receptor 170; 5-oxo-ETE G-protein coupled receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532959 349.00
Check your price
  • OXER1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534061
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
G-protein coupled receptor TG1019; G-protein coupled receptor R527; G-protein coupled receptor 170; 5-oxo-ETE G-protein coupled receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534061 349.00
Check your price
  • OXER1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534062
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (30 µg/ml)
Alternate names:
G-protein coupled receptor TG1019; G-protein coupled receptor R527; G-protein coupled receptor 170; 5-oxo-ETE G-protein coupled receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534062 349.00
Check your price