1 - 30 of 30 products displayed
  • Glucocorticoid Receptor Antibody (Monoclonal, BuGR2)
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Sheep, Rabbit, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA1510
  • Size 100 µg
Applications
Flow Cytometry (1-2 ug/test), Gel Shift (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (5 µg/ml), Immunohistochemistry (PFA fixed) (1/500), Immunoprecipitation (Assay dependent), Western Blot (5 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1510 375.00
Check your price
  • Glucocorticoid Receptor Antibody (Monoclonal, 6E6)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515801
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515801 349.00
Check your price
  • Glucocorticoid Receptor Antibody, FITC (Monoclonal, 5E4)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA181793
  • Size 100 µg
Applications
Flow Cytometry (Neat- 1/10)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181793 650.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, alpha PR-22)
  • Host Mouse
  • Target Species Chicken, Human, Reptile
  • Conjugate Unconjugated
  • Catalog # MA1412
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Gel Shift (Assay Dependent), Immunofluorescence (10 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1412 349.00
Check your price
  • Androgen Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1111B
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1111B 375.00
Check your price
  • Calcitonin Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525594
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
CRT, CT-R, CTR, CTR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525594 349.00
Check your price
  • EEF1A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513540
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Leukocyte receptor cluster member 7; Eukaryotic elongation factor 1 A-1; Elongation factor Tu
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513540 349.00
Check your price
  • EEF1A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513541
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Leukocyte receptor cluster member 7; Eukaryotic elongation factor 1 A-1; Elongation factor Tu
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513541 349.00
Check your price
  • Phospho-INSR (Tyr1158) Antibody (Monoclonal, 8H20L22)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701070
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (1-3 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701070 309.00
Check your price
  • Phospho-IR/IGF1R (Tyr1162, Tyr1163) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44804G
  • Size 10 blots
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44804G 304.00
Check your price
  • INSR beta Antibody (Monoclonal, CT-1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513778
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513778 349.00
Check your price
  • INSR alpha Antibody (Monoclonal, 83-7)
  • Host Mouse
  • Target Species Sheep, Pig, Rabbit, Human, Bovine
  • Conjugate Unconjugated
  • Catalog # AHR0231
  • Size 500 µl
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay-Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (2-4µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0231 305.00
Check your price
  • FAS Antibody, FITC (Monoclonal, LT95)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119604
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119604 225.00
Check your price
  • FAS Antibody, PE (Monoclonal, LT95)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119795
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119795 249.00
Check your price
  • FAS Antibody (Monoclonal, UT-1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119024
  • Size 100 µg
Applications
Flow Cytometry (2-10 ug/ml), Functional Assay (Assay-Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119024 225.00
Check your price
  • FAS Antibody (Monoclonal, LT95)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119311
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119311 199.00
Check your price
  • SRC1 Antibody (Monoclonal, 1135/H4)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA1840
  • Size 100 µL
Applications
ChIP assay (5µl per 10^6 cells), Flow Cytometry (1µg per 10^6 cells), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Steroid receptor coactivator 1; Renal carcinoma antigen NY-REN-52; RIP160; Protein Hin-2; Class E basic helix-loop-helix protein 74; Nuclear receptor coactivator protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1840 275.00
Check your price
  • SRC1 Antibody
  • Host Rabbit
  • Target Species Avian, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1840
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Frozen) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Steroid receptor coactivator 1; Renal carcinoma antigen NY-REN-52; RIP160; Protein Hin-2; Class E basic helix-loop-helix protein 74; Nuclear receptor coactivator protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1840 375.00
Check your price
  • FLT3 Antibody, PE (Monoclonal, 113308)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523648
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Tyrosine-protein kinase receptor flk-2; Fms-like tyrosine kinase 3; Fetal liver kinase 2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523648 339.00
Check your price
  • FLT3 Antibody, APC (Monoclonal, 113308)
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523631
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Tyrosine-protein kinase receptor flk-2; Fms-like tyrosine kinase 3; Fetal liver kinase 2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523631 309.00
Check your price
  • FLT3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514667
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Stem cell tyrosine kinase 1; Fms-like tyrosine kinase 3; Fetal liver kinase-2; FL cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514667 349.00
Check your price
  • CD357 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547883
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Glucocorticoid-induced TNFR-related protein; Activation-inducible TNFR family receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547883 349.00
Check your price
  • FAS Antibody, APC (Monoclonal, LT95)
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA110313
  • Size 100 Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110313 275.00
Check your price
  • FAS Antibody (Monoclonal, 4F8H6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517073
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517073 309.00
Check your price
  • FAS Antibody, Pacific Blue™ (Monoclonal, DX2)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD9528
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD9528 353.00
Check your price
  • FAS Antibody, Pacific Blue™ (Monoclonal, Dx2)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD9528TR
  • Size 25 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD9528TR 135.00
Check your price
  • CD357 Antibody (Monoclonal, YGITR765.4.16)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170089
  • Size 250 ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Glucocorticoid-induced TNFR-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170089 375.00
Check your price
  • AHR Antibody (Monoclonal, RPT9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1513
  • Size 100 µL
Applications
ChIP assay (1:100), Flow Cytometry (1:50), Gel Shift (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:10-1:100), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
Class E basic helix-loop-helix protein 76
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1513 375.00
Check your price
  • TLR4 Antibody (Monoclonal, 76B357.1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA516216
  • Size 100 ug
Applications
Flow Cytometry (0.5-2 µg/10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516216 349.00
Check your price
  • p23 Antibody (Monoclonal, JJ3)
  • Host Mouse
  • Target Species Chicken, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3414
  • Size 100 µL
Applications
Flow Cytometry (1/100), Immunocytochemistry (1:200-1:2000), Immunofluorescence (1:200-1:2000), Immunohistochemistry (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Telomerase-binding protein p23; Progesterone receptor complex p23; Hsp90 co-chaperone; Cytosolic prostaglandin E2 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3414 375.00
Check your price