1 - 4 of 4 products displayed
  • GnRHR Antibody (Monoclonal, GNRH03)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511538
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
gnRH receptor, gnRH-R, gonadotropin-releasing hormone (type 1) receptor 1, leutinizing hormone releasing horomone receptor, leutinizing-releasing hormone receptor, luliberin receptor, type I GnRH receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511538 309.00
Check your price
  • GnRHR Antibody (Monoclonal, A9E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135383
  • Size 100 µg
Applications
ELISA (Assay dependent.), Flow Cytometry (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
GNRHR
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135383 309.00
Check your price
  • GnRHR Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518623
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
GNRHR; LRHR; gonadotropin-releasing hormone receptor; GNRHR1; GRHR; LHRHR; gonadotropin-releasing hormone (type 1) receptor 1; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; luliberin receptor; type I GnRH receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518623 349.00
Check your price
  • GnRHR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533597
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
GNRHR, GnRH receptor, GnRH-R, GRHR, HH7, LHRHR, LRHR, Luliberin receptor, Type I GnRH receptor, GNRHR1, Lh-rh receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533597 349.00
Check your price