1 - 19 of 19 products displayed
  • Tri-Methyl-Histone H3 (Lys27) Antibody (Monoclonal, G.299.10)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511198
  • Size 100 µL
Applications
ChIP assay (1:50), Immunofluorescence (1:1600), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511198 349.00
Check your price
  • Acetyl-Histone H3 (Lys9) Antibody (Monoclonal, J.924.2)
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511195
  • Size 100 µL
Applications
ChIP assay (1:50), Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511195 309.00
Check your price
  • Acetyl-Histone H3 (Lys9, Lys14) Antibody
  • Host Rabbit
  • Target Species C. elegans, Human, Mouse, Non-human primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516194
  • Size 100 µL
Applications
ChIP assay (1:50), Immunofluorescence (1:5000), Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516194 349.00
Check your price
  • Tri-Methyl-Histone H3 (Lys4) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517420
  • Size 100 µL
Applications
ChIP assay (1:100), Immunocytochemistry (1:4000), Immunofluorescence (1:4000), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517420 309.00
Check your price
  • Phospho-Histone H3 (Ser28) Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Hamster, Human, Mouse, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517318
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517318 309.00
Check your price
  • Histone H3 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Fruit fly, Horse, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA516183
  • Size 100 µL
Applications
ChIP assay (1:50), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516183 309.00
Check your price
  • Tri-Methyl-Histone H3 (Lys4) Antibody (Monoclonal, G.532.8)
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA511199
  • Size 100 µL
Applications
ChIP assay (1:50), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511199 309.00
Check your price
  • Histone H3.3 Antibody
  • Host Rabbit
  • Target Species Bovine, Fruit fly, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517697
  • Size 100 µL
Applications
Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517697 275.00
Check your price
  • JMJD6 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523089
  • Size 50 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:2000)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523089 349.00
Check your price
  • DNMT1 Antibody (Monoclonal, 60B1220.1)
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA516169
  • Size 100 ug
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1:10), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (0.1-0.5 µg/ml)
Alternate names:
MCMT; DNA methyltransferase HsaI; CXXC-type zinc finger protein 9; DNA methyltransferase MmuI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516169 349.00
Check your price
  • WDR5 Antibody
  • Host Rabbit
  • Target Species Zebrafish, Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # PA541981
  • Size 100 µl
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:500), Immunoprecipitation (1:500), Western Blot (0.2-1 µg/ml)
Alternate names:
BMP2-induced 3-kb gene protein; WD repeat-containing protein BIG-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541981 309.00
Check your price
  • SMARCB1 Antibody
  • Host Rabbit
  • Target Species Fish, Human, Mouse, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA125450
  • Size 100 µL
Applications
Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
SNF5 homolog; Integrase interactor 1 protein; BRG1-associated factor 47
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125450 375.00
Check your price
  • MSI1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517464
  • Size 100 µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
RNA-binding protein Musashi homolog 1; musashi-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517464 275.00
Check your price
  • beta Catenin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rodent, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519469
  • Size 100 µg
Applications
ELISA (0.1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519469 349.00
Check your price
  • TADA3L Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535154
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Transcriptional adapter 3-like; STAF54; ADA3 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535154 349.00
Check your price
  • SFPQ Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519663
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Polypyrimidine tract-binding protein-associated-splicing factor; DNA-binding p52/p100 complex, 100 kDa subunit; 100 kDa DNA-pairing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519663 349.00
Check your price
  • BRD1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519829
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Bromodomain and PHD finger-containing protein 2; BR140-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519829 349.00
Check your price
  • TAK1 Antibody (Monoclonal, 28H25L68)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700113
  • Size 100 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Transforming growth factor-beta-activated kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700113 303.00
Check your price
  • Phospho-FOXA2 (Thr156) Antibody (Oligoclonal, 10HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710680
  • Size 100 µg
Applications
Immunocytochemistry (3-5 µg/ml), Immunofluorescence (3-5 µg/ml), Western Blot (2-3 µg/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710680 304.00
Check your price