1 - 17 of 17 products displayed
  • HIV1 p24 Antibody (Monoclonal, 2N32 (05-009))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171521
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:500-1:000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 80-90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171521 399.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, 5 (05-001))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171515
  • Size 100 ug
Applications
ELISA (1:64,000), Immunofluorescence (1:1000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 48-62
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171515 399.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, N17 (05-004))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171516
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 C-terminus 165-231
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171516 349.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, 2N34 (05-008))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171520
  • Size 100 ug
Applications
ELISA (1:25000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 74-84
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171520 399.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, G82B)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17040
  • Size 1 mL
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17040 375.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, 2N44 (05-007))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171519
  • Size 100 µg
Applications
ELISA (1:25000), Western Blot (1:5000)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171519 349.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, N29 (05-005))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171517
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171517 349.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, N13 (05-006))
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171518
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171518 349.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, D45F)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17039
  • Size 1 mL
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17039 375.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, 28E0)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183231
  • Size 100 µg
Applications
ELISA (1/20-1/200), Immunofluorescence (1/10-1/50), Immunohistochemistry (Frozen) (1/10-1/50), Western Blot (1/10-1/50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183231 449.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, B59A)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17380
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17380 375.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, C76H)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17379
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17379 375.00
Check your price
  • HIV1 p24 Antibody, FITC (Monoclonal, D45F)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17378
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5-1:20), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17378 375.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, D96A)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17381
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17381 375.00
Check your price
  • HIV1 p24 Antibody (Monoclonal, 1A1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17586
  • Size 100 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17586 309.00
Check your price
  • FIV p24 gag Antibody (Monoclonal, PAK3-2C1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182184
  • Size 250 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Feline Immunodeficiency Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182184 550.00
Check your price
  • FIV p24 gag Antibody (Monoclonal, P24CR1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183937
  • Size 250 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Feline Immunodeficiency Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183937 550.00
Check your price