1 - 30 of 30 products displayed
  • Collagen II Antibody (Monoclonal, 2B1.5)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA137493
  • Size 200 µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml for 30 min @ RT), Western Blot (1-2 µg/ml for 2 hr @ RT)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137493 600.00
Check your price
  • Collagen II Antibody (Monoclonal, 6B3)
  • Host Mouse
  • Target Species Amphibian, Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513026
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513026 309.00
Check your price
  • Collagen II Antibody, Biotin (Monoclonal, 2B1.5)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA512786
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512786 399.00
Check your price
  • Collagen II Antibody (Monoclonal, 2B1.5)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512789
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512789 349.00
Check your price
  • Collagen II Antibody (Monoclonal, CIIC1)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA140065
  • Size 100 µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140065 550.00
Check your price
  • Collagen II Antibody (Monoclonal, M2139)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA140066
  • Size 100 µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140066 550.00
Check your price
  • Collagen III Antibody (Monoclonal, FH-7A)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122147
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (1:4000), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:100-1:1000)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122147 399.00
Check your price
  • Collagen I Antibody (Monoclonal, NFI/20)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183073
  • Size 100 µg
Applications
ELISA (1:1000), Immunohistochemistry (Frozen) (1:20)
Alternate names:
Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183073 349.00
Check your price
  • Collagen IV Antibody (Monoclonal, MC4-HA)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135333
  • Size 200 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (1:20)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135333 425.00
Check your price
  • Procollagen IIA N-Peptide Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0030202
  • Size 200 µl
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0030202 600.00
Check your price
  • Collagen V Antibody (Monoclonal, MC5-HA)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126150
  • Size 500 µL
Applications
ELISA (1:1000), Immunohistochemistry (Frozen) (1:20), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Type V Collagen, Collagen 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126150 425.00
Check your price
  • CD36 Antibody (Monoclonal, 185-1G2)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514112
  • Size 500 µL
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Thrombospondin receptor; Platelet glycoprotein IV; Platelet collagen receptor; PAS-4; PAS IV; Leukocyte differentiation antigen CD36; Glycoprotein IIIb; Fatty acid translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514112 309.00
Check your price
  • MMP2 Antibody (Monoclonal, MMP2/2C1-1D12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1779
  • Size 100 µg
Applications
ELISA (0.1 µg/ml), Immunohistochemistry (Frozen) (5 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:500)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1779 275.00
Check your price
  • MMP2 Antibody (Monoclonal, 2C1-1D12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 351300Z
  • Size 100 µg
Applications
ELISA (0.1 ug/ml), Immunohistochemistry (Frozen) (5 ug/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:500)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
351300Z 304.00
Check your price
  • MMP2 Antibody (Monoclonal, 101)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rabbit, Rhesus monkey, Rat
  • Conjugate Unconjugated
  • Catalog # 436000
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase; TBE-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
436000 309.00
Check your price
  • MMP2 Antibody (Monoclonal, MMP2/2C1)
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116640
  • Size 100 µL
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (2-5 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116640 349.00
Check your price
  • MMP2 Antibody (Monoclonal, MMP2/8B4)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116641
  • Size 100 µL
Applications
ELISA (0.03 µg/ml), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (2 µg/ml), Immunohistochemistry (Paraffin) (2 µg/ml), Immunoprecipitation (1-2 µg/100-500 µg total protein), Western Blot (1-2 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116641 375.00
Check your price
  • MMP2 Antibody (Monoclonal, CA-4001 or CA719E3C)
  • Host Mouse
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513590
  • Size 500 µL
Applications
Inhibition Assays (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513590 349.00
Check your price
  • CD36 Antibody (Monoclonal, D-2712)
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA146000
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Immunoprecipitation (10-20 µg), Western Blot (1:500)
Alternate names:
Platelet glycoprotein IV; PAS-4; PAS IV; Glycoprotein IIIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146000 309.00
Check your price
  • CD36 Antibody (Monoclonal, SMØ)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516941
  • Size 1 mg
Applications
Flow Cytometry (1:25-1:50), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Thrombospondin receptor; Platelet glycoprotein IV; Platelet collagen receptor; PAS-4; PAS IV; Leukocyte differentiation antigen CD36; Glycoprotein IIIb; Fatty acid translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516941 900.00
Check your price
  • MMP2 Antibody (Monoclonal, 4D3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112892
  • Size 200 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112892 349.00
Check your price
  • Collagen X Antibody, Biotin (Monoclonal, X-AC9)
  • Host Mouse
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # MA514265
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2-4 µg/ml)
Alternate names:
collagen, type X, alpha 1, procollagen, type X, alpha 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514265 600.00
Check your price
  • Collagen X Antibody (Monoclonal, X-AC9)
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # MA514268
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2-4 µg/ml)
Alternate names:
collagen, type X, alpha 1, procollagen, type X, alpha 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514268 475.00
Check your price
  • CD105 Antibody, Biotin (Monoclonal, MJ7/18)
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517944
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517944 129.00
Check your price
  • CD105 Antibody, FITC (Monoclonal, MJ7/18)
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517945
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517945 129.00
Check your price
  • CD105 Antibody, PE (Monoclonal, MJ7/18)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517946
  • Size 50 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517946 129.00
Check your price
  • CD105 Antibody (Monoclonal, MJ7/18)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517943
  • Size 250 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517943 225.00
Check your price
  • MMP13 Antibody (Monoclonal, 3H13L17)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701287
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-3µg/ml)
Alternate names:
Matrix metalloproteinase-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
701287 309.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, TB21)
  • Host Mouse
  • Target Species Bovine, Human, Mustelid, Mink, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516949
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta,TGF-beta 1 protein; TGF-beta-1; latency-associated peptide; transforming growth factor beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516949 375.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, TB21)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518023
  • Size 100 µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (5-20 ng/mL)
Alternate names:
CED; DPD1; TGFB; TGFbeta; TGFB1; Latency-associated peptide; LAP; Transforming growth factor beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518023 425.00
Check your price