1 - 30 of 30 products displayed
  • Phospho-PKC lambda/iota (Thr555, Thr563) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44968G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (Assay Dependent)
Alternate names:
nPKC-iota; Atypical protein kinase C-lambda/iota
Catalog # Size Price (AUD) Your Price (AUD) Qty
44968G 304.00
Check your price
  • Non-Phospho PTEN (Ser380+Thr382+Thr383) Antibody Polyclonal (PA5-17153)
  • Host Rabbit
  • Target Species Mouse, Rat, Human, Chicken
  • Conjugate
  • Catalog # PA517153
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517153 309.00
Check your price
  • Phospho-PTEN (Ser385) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441064G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441064G 309.00
Check your price
  • Phospho-Cdc25C (Ser216) Antibody (Monoclonal, K.827.0)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515146
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Dual specificity phosphatase Cdc25C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515146 309.00
Check your price
  • Phospho-PTPN3 (Ser459) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA535413
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein-tyrosine phosphatase H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535413 349.00
Check your price
  • Phospho-Tau (Ser422) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OPA103151
  • Size 100 µL
Applications
Western Blot (Assay-Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103151 349.00
Check your price
  • Phospho-Tau (Ser422) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44764G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44764G 309.00
Check your price
  • Aprataxin Antibody (Monoclonal, 2H6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522070
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Forkhead-associated domain histidine triad-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522070 309.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody (Monoclonal, 22B1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • Phospho-ATRIP (Ser224) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16678
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
ATM and Rad3-related-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16678 309.00
Check your price
  • TNF alpha Antibody (Monoclonal, 4F11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1410802
  • Size 200 µl
Applications
ELISA (1:16000)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1410802 425.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody (Oligoclonal, B10HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710088
  • Size 100 µg
Applications
ChIP assay (1 ul), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1-5 ug/ml)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710088 309.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody (Monoclonal, B10H8L5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701057
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.1-2 ug/ml)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701057 309.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 44674G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
44674G 309.00
Check your price
  • Phospho-p70 S6 Kinase (Thr389) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44920G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44920G 309.00
Check your price
  • Phospho-AMPK alpha-1,2 (Ser485, Ser491) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517543
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517543 309.00
Check your price
  • Phospho-AMPK alpha-1 (Ser485) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517542
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Tau-protein kinase PRKAA1; Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517542 309.00
Check your price
  • Phospho-MAPKAPK2 (Thr334) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44516G
  • Size 10 blots
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Western Blot (Assay Dependent)
Alternate names:
MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44516G 309.00
Check your price
  • Phospho-GSK3 alpha/beta (Tyr279, Tyr216) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # OPA103083
  • Size 100 µL
Applications
Western Blot (Assay-Dependent)
Alternate names:
Serine/threonine-protein kinase GSK3B; Serine/threonine-protein kinase GSK3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103083 375.00
Check your price
  • Phospho-4E-BP1 (Ser65) Antibody (Monoclonal, T.730.9)
  • Host Rabbit
  • Target Species Chicken, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514948
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Phosphorylated heat- and acid-stable protein regulated by insulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514948 309.00
Check your price
  • Phospho-IkB alpha (Ser32) Antibody (Oligoclonal)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 710295
  • Size 100 µg
Applications
Flow Cytometry (1-3µg x 10^6 cells), Western Blot (1-3µg/ml)
Alternate names:
Major histocompatibility complex enhancer-binding protein MAD3; I-kappa-B-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
710295 309.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA112736
  • Size 10 Miniblots
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
Myelin xP42 protein kinase; Myelin basic protein kinase; M phase MAP kinase; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112736 449.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204) Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 368800
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:500), Western Blot (1-3 µg/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
368800 308.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112741
  • Size 10 Miniblots
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112741 399.00
Check your price
  • Phospho-CDK9 (Thr186) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517197
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Cell division protein kinase 9; Tat-associated kinase complex catalytic subunit; Serine/threonine-protein kinase PITALRE; Cell division cycle 2-like protein kinase 4; C-2K
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517197 309.00
Check your price
  • Phospho-PAK1/2/3 (Thr402) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14636
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p68-PAK; p21-activated kinase 1; Protein kinase MUK2; Alpha-PAK; p65-PAK; p21-activated kinase 3; Beta-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14636 375.00
Check your price
  • Phospho-ZAP70/Syk (Tyr319, Tyr352) Antibody (Monoclonal, R.658.6)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Hamster, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA515095
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Spleen tyrosine kinase; p72-Syk; Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein; p72Syk
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515095 309.00
Check your price
  • Phospho-GSK3 alpha/beta (Ser9, Ser21) Antibody (Monoclonal, K.857.1)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515023
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase GSK3A; Factor A; Serine/threonine-protein kinase GSK3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515023 309.00
Check your price
  • Nonphospho-4E-BP1 (Thr46) Antibody (Monoclonal, E.638.3)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515032
  • Size 100 µL
Applications
Flow Cytometry (1:200), Western Blot (1:1000)
Alternate names:
Phosphorylated heat- and acid-stable protein regulated by insulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515032 309.00
Check your price
  • Phospho-PKC beta-1,2 (Thr500) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44955G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Protein kinase C beta type, PKC-B, PKC-beta, PRKCB, PKCB, PRKCB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44955G 309.00
Check your price