1 - 5 of 5 products displayed
  • PMCA ATPase Antibody (Monoclonal, 5F10)
  • Host Mouse
  • Target Species Amphibian, Dog, Chicken, Cat, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3914
  • Size 100 µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin, Frozen) (1:500), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Plasma membrane calcium pump isoform 1; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium ATPase isoform 4; Matrix-remodeling-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3914 375.00
Check your price
  • ATP1B1 Antibody (Monoclonal, M17-P5-F11)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # MA3930
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:1000), Immunohistochemistry (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:10,000)
Alternate names:
Sodium/potassium-dependent ATPase subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3930 375.00
Check your price
  • ATP1A1 Antibody (Monoclonal, M7-PB-E9)
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3928
  • Size 400 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:20), Immunohistochemistry (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3928 375.00
Check your price
  • Phospho-ATP1A1 (Tyr10) Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517061
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517061 309.00
Check your price
  • ATP1A1 Antibody (Monoclonal, 9-A5)
  • Host Mouse
  • Target Species Dog, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA3924
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1/50), Immunocytochemistry (Assay dependent), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:1,000), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/100), Immunoprecipitation (Assay dependent), Inhibition Assays (Assay De
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3924 375.00
Check your price