1 - 7 of 7 products displayed
  • gp210 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA129888
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Pore membrane protein of 210 kDa; Nucleoporin Nup210; Nuclear envelope pore membrane protein POM 210
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129888 399.00
Check your price
  • gp210 Antibody
  • Host Rabbit
  • Target Species Mammal, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA15781
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Pore membrane protein of 210 kDa; Nucleoporin Nup210; Nuclear envelope pore membrane protein POM 210
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15781 475.00
Check your price
  • gp210 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA15782
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Pore membrane protein of 210 kDa; Nucleoporin Nup210; Nuclear envelope pore membrane protein POM 210
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15782 425.00
Check your price
  • POM121 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536498
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p145; Pore membrane protein of 121 kDa; Nucleoporin Nup121; Nuclear envelope pore membrane protein POM 121A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536498 309.00
Check your price
  • POM121 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539308
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p145; Pore membrane protein of 121 kDa; Nucleoporin Nup121; Nuclear envelope pore membrane protein POM 121A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539308 309.00
Check your price
  • gp210 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526961
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Pore membrane protein of 210 kDa; Nucleoporin Nup210; Nuclear envelope pore membrane protein POM 210
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526961 349.00
Check your price
  • POM121 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA527623
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Pore membrane protein of 121 kDa; Nucleoporin Nup121; Nuclear envelope pore membrane protein POM 121A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527623 349.00
Check your price