1 - 5 of 5 products displayed
  • Nesprin 2 Antibody (Monoclonal, K20-478-5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518075
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunomicroscopy (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:2500)
Alternate names:
Synaptic nuclear envelope protein 2; Nucleus and actin connecting element protein; Nuclear envelope spectrin repeat protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518075 425.00
Check your price
  • Nesprin 1 Antibody (Monoclonal, MANNES1A(7A12))
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA518077
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:400-1:800)
Alternate names:
Synaptic nuclear envelope protein 1; Nuclear envelope spectrin repeat protein 1; Myocyte nuclear envelope protein 1; Enaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518077 425.00
Check your price
  • Nesprin 1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA15839
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:500)
Alternate names:
Synaptic nuclear envelope protein 1; Nuclear envelope spectrin repeat protein 1; Myocyte nuclear envelope protein 1; Enaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15839 475.00
Check your price
  • SYNE4 Antibody
  • Host Rabbit
  • Target Species Bovine, Horse, Human
  • Conjugate Unconjugated
  • Catalog # PA542662
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Nuclear envelope spectrin repeat protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542662 309.00
Check your price
  • SYNE4 Antibody
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA542661
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Nuclear envelope spectrin repeat protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542661 309.00
Check your price