1 - 5 of 5 products displayed
  • Phospho-CaMKII alpha/delta (Thr286) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536838
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536838 349.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody (Monoclonal, 22B1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126727
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
CAMKA, KIAA0968
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126727 399.00
Check your price
  • Phospho-CaMKII alpha/beta (Thr286) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14614
  • Size 100 µL
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
phospho-Ca2+/Calmodulin-Dependent Protein Kinase II (Thr286)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14614 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 44674G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
44674G 309.00
Check your price