1 - 3 of 3 products displayed
  • PSD95 Antibody (Monoclonal, 7E3-1B8)
  • Host Mouse
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA1046
  • Size 100 µL
Applications
Flow Cytometry (2 ug/test), Immunocytochemistry (1:500), Immunofluorescence (1:200), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1046 375.00
Check your price
  • PSD95 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14667
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14667 375.00
Check your price
  • PARD6B Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542264
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Par-6 partitioning defective 6 homolog beta (C. elegans), PAR6B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542264 309.00
Check your price