1 - 2 of 2 products displayed
  • ZMPSTE24 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA116964
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1.0-2 µg/ml)
Alternate names:
Zinc metalloproteinase Ste24 homolog; Prenyl protein-specific endoprotease 1; Farnesylated proteins-converting enzyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116964 349.00
Check your price
  • ZMPSTE24 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116965
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Zinc metalloproteinase Ste24 homolog; Prenyl protein-specific endoprotease 1; Farnesylated proteins-converting enzyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116965 309.00
Check your price