1 - 9 of 9 products displayed
  • Rabies Virus Antibody (Monoclonal, 5B12)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA120006
  • Size 100 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120006 309.00
Check your price
  • Rabies Virus Antibody (Monoclonal, 4G4)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA120005
  • Size 100 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120005 349.00
Check your price
  • Rabies Virus Antibody (Monoclonal)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA121549
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121549 349.00
Check your price
  • Rabies Virus Antibody (Monoclonal, C94A)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17079
  • Size 1 mL
Applications
ELISA (1:20-1:200), Immunocytochemistry (1:10-50), Immunofluorescence (1:10-50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17079 349.00
Check your price
  • Rabies Virus Antibody (Monoclonal, 0.N.541)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA125788
  • Size 50 µg
Applications
ELISA (Assay Dependent), Inhibition Assays (Assay Dependent), Neutralization (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125788 375.00
Check your price
  • Rabies Virus Antibody (Monoclonal, Rab-50)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA513572
  • Size 500 µL
Applications
ELISA (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513572 309.00
Check your price
  • Rabies Virus Antibody (Monoclonal, RV1C5)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190618
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190618 249.00
Check your price
  • Rabies Virus Antibody (Monoclonal, RAB-50)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA135778
  • Size 100 µg
Applications
Blocking Assay (Assay Dependent), ELISA (1:50), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135778 309.00
Check your price
  • Rabies Virus Antibody (Monoclonal, 1C5)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516456
  • Size 200 ug
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516456 399.00
Check your price