1 - 30 of 30 products displayed
  • Rat anti-Mouse IgG (Kappa light chain)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # SA125258
  • Size 500 µg
Applications
Flow Cytometry (1:100 (10 µl for 10^6 cells in 100 µl))
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125258 483.00
Check your price
  • Rat anti-Mouse IgG (Kappa light chain), HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA134732
  • Size 250 µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134732 472.00
Check your price
  • Rat anti-Mouse IgG (Heavy chain), Biotin
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA125198
  • Size 125 µg
Applications
ELISA (4-6 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125198 197.00
Check your price
  • Rat anti-Mouse IgG (Heavy chain)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA180696
  • Size 250 µg
Applications
ELISA (1.25 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180696 402.00
Check your price
  • Rat anti-Mouse IgG3 (Heavy chain)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # SA125253
  • Size 200 µg
Applications
ELISA (5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125253 225.00
Check your price
  • Rat anti-Mouse IgG, HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 046020
  • Size 1 mL
Applications
ELISA (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (Paraffin) (1:500-1:1,000), Immunoprecipitation (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046020 199.00
Check your price
  • Rat anti-Mouse IgG, Biotin
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 046040
  • Size 1 mL
Applications
ELISA (1:1,000), Immunofluorescence (1:1,000), Immunohistochemistry (Paraffin) (1:1,000), Immunoprecipitation (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046040 179.00
Check your price
  • Rat anti-Mouse IgG, HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # SA135636
  • Size 1 mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135636 1
Check your price
  • Rat anti-Mouse IgG
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # SA135633
  • Size 1 mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135633 806.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 680
  • Host Donkey
  • Target Species Mouse
  • Conjugate DyLight® 680
  • Catalog # SA510170
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510170 157.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 800
  • Host Rabbit
  • Target Species Mouse
  • Conjugate DyLight® 800
  • Catalog # SA510164
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510164 156.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 680
  • Host Rabbit
  • Target Species Mouse
  • Conjugate DyLight® 680
  • Catalog # SA510162
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510162 160.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 488
  • Host Rabbit
  • Target Species Mouse
  • Conjugate DyLight® 488
  • Catalog # SA510158
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510158 156.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 594
  • Host Donkey
  • Target Species Mouse
  • Conjugate DyLight® 594
  • Catalog # SA510168
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510168 157.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 550
  • Host Donkey
  • Target Species Mouse
  • Conjugate DyLight® 550
  • Catalog # SA510167
  • Size 500 ug
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (1:50-1:500), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510167 158.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 650
  • Host Rabbit
  • Target Species Mouse
  • Conjugate DyLight® 650
  • Catalog # SA510161
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510161 158.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 550
  • Host Rabbit
  • Target Species Mouse
  • Conjugate DyLight® 550
  • Catalog # SA510159
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510159 156.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 800
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 800
  • Catalog # SA510176
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510176 157.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 650
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 650
  • Catalog # SA510174
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510174 156.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed, DyLight® 550
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 550
  • Catalog # SA510173
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510173 156.00
Check your price
  • Donkey anti-Mouse IgG, HRP
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA128748
  • Size 100 µg
Applications
Dot blot (Assay dependent), ELISA (1:5000-1:20,000), Immunohistochemistry (Paraffin, Frozen) (1:1000-1:5000), Immunomicroscopy (Assay dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128748 149.00
Check your price
  • Rat anti-Mouse IgG2a (Heavy chain), HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA516790
  • Size 500 ug
Applications
ELISA (0.5 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516790 700.00
Check your price
  • Rat anti-Mouse IgG1 (Heavy chain), HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA516786
  • Size 500 ug
Applications
ELISA (500 ng/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516786 650.00
Check your price
  • Rat anti-Mouse IgG1, FITC
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # RMG101
  • Size 500 µL
Applications
Flow Cytometry (0.5-2 µg/mL), Immunofluorescence (0.5-2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMG101 123.00
Check your price
  • Rat anti-Mouse IgM Heavy Chain, Sepharose® 4B
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # 046841
  • Size 2mL
Applications
ELISA (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046841 392.00
Check your price
  • Rat anti-Mouse IgM Heavy Chain, HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 046820
  • Size 1 mL
Applications
ELISA (1:500-1:1,000), Flow Cytometry (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046820 199.00
Check your price
  • Rat anti-Mouse IgM Heavy Chain
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 046800
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046800 193.00
Check your price
  • Rat anti-Mouse IgG2a, Biotin
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 046240
  • Size 1 mL
Applications
ELISA (1:1,000), Flow Cytometry (1:1,000), Immunofluorescence (1:1,000), Immunohistochemistry (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046240 179.00
Check your price
  • Rat anti-Mouse IgG2a, HRP
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 046220
  • Size 1 mL
Applications
ELISA (1:500-1:1,000), Flow Cytometry (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046220 199.00
Check your price
  • Rat anti-Mouse IgG2a
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 046200
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
046200 192.00
Check your price