1 - 3 of 3 products displayed
  • SLC5A5 Antibody (Monoclonal, FP5A)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512308
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400-800), Western Blot (Assay Dependent)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512308 349.00
Check your price
  • SLC5A5 Antibody (Monoclonal, SPM186)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121239
  • Size 500 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1/50), Western Blot (1-2 µg/ml.)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121239 425.00
Check your price
  • SLC5A5 Antibody
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA136151
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:1,200-1:2,000)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136151 900.00
Check your price