1 - 30 of 30 products displayed
  • Sodium/Calcium Exchanger Antibody (Monoclonal, C2C12)
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3926
  • Size 100 µL
Applications
Flow Cytometry (1/100), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:20-50), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/100), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Solute carrier family 8 member 1; Na(+)/Ca(2+)-exchange protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3926 375.00
Check your price
  • SLC5A5 Antibody (Monoclonal, SPM186)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121239
  • Size 500 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1/50), Western Blot (1-2 µg/ml.)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121239 425.00
Check your price
  • SLC5A5 Antibody (Monoclonal, FP5A)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512308
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400-800), Western Blot (Assay Dependent)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512308 349.00
Check your price
  • SLC5A5 Antibody
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA136151
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:1,200-1:2,000)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136151 900.00
Check your price
  • SLC24A2 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA1926
  • Size 200 µL
Applications
Immunohistochemistry (Frozen) (1:100), Western Blot (1:100)
Alternate names:
Solute carrier family 24 member 2; Retinal cone Na-Ca+K exchanger; Na(+)/K(+)/Ca(2+)-exchange protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1926 275.00
Check your price
  • SLCO1C1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # PA542469
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Thyroxine transporter; Solute carrier family 21 member 14; Organic anion-transporting polypeptide 14; Organic anion transporter polypeptide-related protein 5; Organic anion transporter F; Organic anion transporter 2; Blood-brain barrier-specific anion transporter 1; BBB-specific anion transporter type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542469 309.00
Check your price
  • NTCP Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542101
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 10 member 1; Sodium/taurocholate cotransporting polypeptide; Na(+)/taurocholate transport protein; Na(+)/bile acid cotransporter; Cell growth-inhibiting gene 29 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542101 309.00
Check your price
  • NTCP Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525614
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Solute carrier family 10 member 1; Sodium/taurocholate cotransporting polypeptide; Na(+)/taurocholate transport protein; Na(+)/bile acid cotransporter; Cell growth-inhibiting gene 29 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525614 349.00
Check your price
  • SLC20A2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542142
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 20 member 2; Phosphate transporter 2; Gibbon ape leukemia virus receptor 2; Type III sodium-dependent phosphate transporter; Receptor for amphotropic viruses 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542142 309.00
Check your price
  • SLC6A15 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536502
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
sodium- and chloride-dependent neurotransmitter transporter NTT73; Transporter v7-3; Solute carrier family 6 member 15; Sodium-coupled branched-chain amino-acid transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536502 309.00
Check your price
  • SLC6A15 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA539325
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
sodium- and chloride-dependent neurotransmitter transporter NTT73; Transporter v7-3; Solute carrier family 6 member 15; Sodium-coupled branched-chain amino-acid transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539325 309.00
Check your price
  • SLC4A8/SLC4A10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539473
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Solute carrier family 4 member 10; k-NBC3; Solute carrier family 4 member 8; Electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral Na(+)-driven Cl-HCO3 exchanger
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539473 309.00
Check your price
  • EAAC1 Antibody (Monoclonal, 35-A9)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 321000
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (1-5 ug), Western Blot (1-5 ug/ml)
Alternate names:
Solute carrier family 1 member 1; Sodium-dependent glutamate/aspartate transporter 3; Neuronal and epithelial glutamate transporter; Excitatory amino-acid carrier 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
321000 309.00
Check your price
  • SLC23A2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542447
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Sodium-dependent vitamin C transporter 2; Na(+)/L-ascorbic acid transporter 2; Yolk sac permease-like molecule 2; Nucleobase transporter-like 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542447 309.00
Check your price
  • OATP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526615
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Solute carrier family 21 member 6; Sodium-independent organic anion-transporting polypeptide 2; OATP-C; Liver-specific organic anion transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526615 349.00
Check your price
  • OATP2/OATP8 Antibody (Monoclonal, MDQ)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3935
  • Size 500 µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Frozen) (1:20), Immunoprecipitation (Assay dependent)
Alternate names:
Solute carrier family 21 member 8; Organic anion-transporting polypeptide 8; Organic anion transporter 8; Liver-specific organic anion transporter 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3935 375.00
Check your price
  • SLC4A7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525563
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Solute carrier family 4 member 7; Sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 2; Electroneutral Na/HCO(3) cotransporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525563 349.00
Check your price
  • OATP2 Antibody (Monoclonal, ESL)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3934
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Solute carrier family 21 member 6; Sodium-independent organic anion-transporting polypeptide 2; OATP-C; Liver-specific organic anion transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3934 375.00
Check your price
  • SLC20A1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542448
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 20 member 1; Phosphate transporter 1; Leukemia virus receptor 1 homolog; Gibbon ape leukemia virus receptor 1; RPHO-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542448 309.00
Check your price
  • OATP1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542445
  • Size 100 µl
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 21 member 3; Sodium-independent organic anion transporter; Organic anion-transporting polypeptide 1; OATP-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542445 309.00
Check your price
  • OATP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA536610
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Solute carrier family 21 member 3; Sodium-independent organic anion transporter; Organic anion-transporting polypeptide 1; OATP-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536610 309.00
Check your price
  • SLC4A11 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519207
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Solute carrier family 4 member 11; Sodium borate cotransporter 1; Bicarbonate transporter-related protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519207 349.00
Check your price
  • SLC12A1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541995
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Kidney-specific Na-K-Cl symporter; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; BSC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541995 309.00
Check your price
  • CD98 Antibody, PE (Monoclonal, MEM-108)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110314
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110314 309.00
Check your price
  • CD98 Antibody (Monoclonal, 44D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516514
  • Size 200 ug
Applications
Flow Cytometry (1:50-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516514 375.00
Check your price
  • CD98 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185012
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (15-25 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185012 650.00
Check your price
  • SLC24A2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537224
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Solute carrier family 24 member 2; Retinal cone Na-Ca+K exchanger; Na(+)/K(+)/Ca(2+)-exchange protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537224 309.00
Check your price
  • NKCC1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533024
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (7.5-10 µg/ml)
Alternate names:
Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Basolateral Na-K-Cl symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533024 349.00
Check your price
  • NKCC1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533023
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (7.5-10 µg/ml)
Alternate names:
Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Basolateral Na-K-Cl symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533023 349.00
Check your price
  • NKCC1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534196
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml)
Alternate names:
Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Basolateral Na-K-Cl symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534196 349.00
Check your price