1 - 3 of 3 products displayed
  • NTCP Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542101
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 10 member 1; Sodium/taurocholate cotransporting polypeptide; Na(+)/taurocholate transport protein; Na(+)/bile acid cotransporter; Cell growth-inhibiting gene 29 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542101 309.00
Check your price
  • NTCP Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525614
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Solute carrier family 10 member 1; Sodium/taurocholate cotransporting polypeptide; Na(+)/taurocholate transport protein; Na(+)/bile acid cotransporter; Cell growth-inhibiting gene 29 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525614 349.00
Check your price
  • SLC10A2 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518990
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Solute carrier family 10 member 2; Sodium/taurocholate cotransporting polypeptide, ileal; Na(+)-dependent ileal bile acid transporter; Ileal sodium-dependent bile acid transporter; Ileal Na(+)/bile acid cotransporter; Apical sodium-dependent bile acid transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518990 349.00
Check your price