1 - 5 of 5 products displayed
  • Furin Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1062
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (1:100), Western Blot (1:1,000)
Alternate names:
Paired basic amino acid residue-cleaving enzyme; Dibasic-processing enzyme; Prohormone convertase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1062 375.00
Check your price
  • ACTH Antibody (Monoclonal, AH26)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513455
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (0.2-0.4 µg/mL)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513455 309.00
Check your price
  • POMC Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA185109
  • Size 500 µL
Applications
ELISA (1/500-1/1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1/100-1/200)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185109 375.00
Check your price
  • POMC Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518368
  • Size 100 µg
Applications
Immunohistochemistry (4-6 µg/ml), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518368 349.00
Check your price
  • POMC Antibody (Monoclonal, 6Y8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518042
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1-4 ug/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518042 399.00
Check your price