1 - 2 of 2 products displayed
  • C. tetani TeNT Light Chain Antibody (Monoclonal, 604023)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA524273
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
TeNT-LC, Tentoxylysin, tetX, Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524273 275.00
Check your price
  • Clostridium tetani Tetanus Toxin Antibody (Monoclonal, TetH6)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA174231
  • Size 200 µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C. tetani, pE88_60
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174231 399.00
Check your price