1 - 2 of 2 products displayed
  • Vomeronasal 1 Receptor A1/A13/A14 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # OSR00230W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Vomeronasal type-1 receptor A1; Vomeronasal type-1 receptor 51; Pheromone receptor VN1; Vomeronasal type-1 receptor A13
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00230W 309.00
Check your price
  • Vomeronasal 1 Receptor B8/A13 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00231W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:3000), Western Blot (1:300-1:3000)
Alternate names:
Vomeronasal type-1 receptor B8; Vomeronasal type-1 receptor A13
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00231W 309.00
Check your price