1 - 10 of 10 products displayed
  • Alcohol Dehydrogenase Antibody, Biotin
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Biotin
  • Catalog # PA127272
  • Size 1 mg
Applications
Dot blot (Assay Dependent), ELISA (1:1000-1:5000), Immunomicroscopy (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
YADH-1; Alcohol dehydrogenase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127272 309.00
Check your price
  • ATF1 Antibody (Monoclonal, ATF1 2A9/8)
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA121313
  • Size 100 µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
EWS-ATF1, FUS/ATF-1, TREB36
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121313 399.00
Check your price
  • ERAB Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532392
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type II HADH; Short-chain type dehydrogenase/reductase XH98G2; Short chain dehydrogenase/reductase family 5C member 1; Mitochondrial ribonuclease P protein 2; Endoplasmic reticulum-associated amyloid beta-peptide-binding protein; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 17-beta-hydroxysteroid dehydrogenase 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532392 349.00
Check your price
  • Hexokinase Antibody, HRP
  • Host Rabbit
  • Target Species Yeast
  • Conjugate HRP
  • Catalog # PA127232
  • Size 1 mg
Applications
ELISA (1:4000-1:20,000), Western Blot (1:500-1:2000)
Alternate names:
Hexokinase-A; Hexokinase PI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127232 349.00
Check your price
  • ATM Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522689
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522689 375.00
Check your price
  • ATM Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522692
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522692 375.00
Check your price
  • ATM Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522690
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522690 375.00
Check your price
  • ATM Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522400
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522400 375.00
Check your price
  • ATM Antibody, HRP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522691
  • Size 50 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522691 375.00
Check your price
  • ATM Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116503
  • Size 50µL
Applications
ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116503 Not Orderable Online
Check your price