1 - 3 of 3 products displayed
  • ZNF7 Antibody (Monoclonal, 6F2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520242
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein KOX4; Zinc finger protein HF.16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520242 325.00
Check your price
  • ZNF7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521521
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger protein KOX4; Zinc finger protein HF.16
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521521 349.00
Check your price
  • ZNF7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538583
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Zinc finger protein KOX4; Zinc finger protein HF.16
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538583 309.00
Check your price