1 - 3 of 3 products displayed
  • ZNF24 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540731
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Zinc finger protein ZF-12; Zinc finger protein 191; Hypomyelinated CNS protein; Zinc finger protein KOX17; Zinc finger and SCAN domain-containing protein 3; Retinoic acid suppression protein A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540731 309.00
Check your price
  • ZNF24 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538582
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Zinc finger protein ZF-12; Zinc finger protein 191; Hypomyelinated CNS protein; Zinc finger protein KOX17; Zinc finger and SCAN domain-containing protein 3; Retinoic acid suppression protein A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538582 309.00
Check your price
  • ZNF24 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547521
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Zinc finger protein KOX17; Zinc finger protein 191; Zinc finger and SCAN domain-containing protein 3; Retinoic acid suppression protein A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547521 349.00
Check your price