Medio Advanced RPMI 1640
Gibco™
Medio Advanced RPMI 1640
Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cellsMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
12633012500 mL
1263302010 x 500 mL
Número de catálogo 12633012
Precio (MXN)
-
Cantidad:
500 mL
Customize this product
Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic RPMI 1640, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced RPMI 1640, with no adaptation, include Sp2, Vero, Raji, Daudi, and Jurkat.
This Advanced RPMI 1640 is manufactured as follows:
With: glucose, non-essential amino acids, sodium pyruvate, phenol red
Without: L-glutamine, HEPES
The complete formulation is available.
Gibco Advanced RPMI 1640 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.
Advanced RPMI 1640 requires supplementation with 1-5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX supplement. Many cell lines do not require adaptation to this medium. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced RPMI 1640 uses a sodium bicarbonate buffer system (2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
iAdvanced Media: Reduced Sera Solutions. Quest, 2004. 1.3:26-30.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Línea de célulasSp2, Vero, Raji, Daudi y Jurkat
Concentración1 X
Calidad de fabricacióncGMP-compliant under the ISO 13485 standard
Línea de productosGibco
Tipo de productoMedio RPMI 1640 (medio Roswell Park Memorial Institute 1640)
Cantidad500 mL
Duración de almacenamiento12 meses a partir de la fecha de fabricación
Condiciones de envíoTemperatura ambiente
ClasificaciónOrigen animal
FormularioLíquido
Serum LevelSuero reducido
EsterilidadEstéril con filtro
Sterilization MethodEstéril con filtro
Con aditivosGlucosa, Rojo de fenol, Piruvato sódico, Aminoácidos no esenciales
Sin aditivosSin glutamina, Sin HEPES
Unit SizeEach
Contenido y almacenamiento
Condiciones de almacenamiento: De 2 a 8 °C. Proteger de la luz
Condiciones de envío: Ambiente
Vida útil: 12 meses a partir de la fecha de fabricación
Preguntas frecuentes
What is the glucose concentration in Advanced RPMI 1640 Medium (Cat. No. 12633012, 12633020)?
Advanced RPMI 1640 Medium (Cat. No. 12633012, 12633020) contains glucose (D-Glucose or Dextrose) at a concentration of 2 g/L.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer Advanced RPMI 1640 Medium (Cat. No. 12633012 or 12633020) without phenol red?
We do not offer Advanced RPMI 1640 medium without phenol red as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Advanced RPMI 1640 Medium and other RPMI 1640 formulations?
Advanced Media are widely used basal media, which require 50-90% less Fetal Bovine Serum (FBS) supplementation compared to conventional basal media formulations. The Advanced media is unique due to the addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How long can I keep my media after supplementing with serum?
Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?
We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.