Food allergies affect approximately 250 million people worldwide.1 Understanding a food allergy diagnosis is vital for making confident food choices and managing symptoms that can vary from mild to severe. This page provides valuable information on food allergies including causes, symptoms, diagnosis and testing options.
Food allergies are a type of allergic reaction that occurs when the immune system reacts to typically harmless substances, like whole allergens from milk, eggs, fish, or peanuts. It’s also possible to have allergic reactions to specific molecules, called allergen components, within whole allergens.
Food allergies can cause a wide variety of symptoms from less severe reactions like rashes, stomach issues and itchy mouth, to more severe reactions like anaphylaxis.
1 in 10 adults2
1 in 13 children3
Food intolerances, like lactose intolerance and wheat sensitivity, mimic food allergy symptoms but are not caused by an immune system reaction.4 They result from enzyme deficiencies or pharmacological reactions. Food allergies are solely immune system responses.
A food allergy develops when your body’s immune system has an abnormal response to a specific food and creates IgE antibodies.5
The next time you eat that specific food, the specific IgE antibodies cause an immune response, resulting in food allergy symptoms.5
10% of adults, globally, have a food allergy.6
More than 50% of adults with food allergies have experienced a severe reaction.8
80% of children outgrow milk, egg, soy, and wheat allergies.7
More than 40% of children with food allergies have experienced a severe reaction.3
90% of food allergies are caused by nine common allergens.9
Because a large population of people react to these 9 allergens, they are deemed major allergens and must be called out on food labels.
5 additional common food allergens include: celery, mustard, sulphur dioxide and sulphites, lupin and molluscs.
Pollen food allergy syndrome, or oral allergy syndrome (OAS), occurs when someone with a pollen allergy eats a specific fruit or vegetable—like kiwi, celery, tomatoes, peppers, and avocado—and experiences symptoms like itchy mouth and throat.
Food allergy symptoms vary from stomach cramps and an itchy mouth to rashes and fatigue. While anaphylaxis is the most severe reaction, food allergies can also cause minor reactions depending on the individual.
The only way to be clinically diagnosed with a food allergy is to have a medical history of reactions paired with a positive allergy test.10 Serologic food allergy testing evaluates specific IgE antibodies (sIgE) found in the blood, which, at high levels, indicates what food allergens may be causing you to react.
Healthcare providers use two main types of food allergy testing: specific IgE blood testing and skin prick testing (SPT). SPT is usually only offered by an allergist; however, specific IgE blood testing can be used by a wide range of clinicians, such as primary care providers, urgent care clinicians, pediatricians and internists.
Be careful not to confuse at-home food sensitivity or food intolerance testing as allergy testing. These for-purchase testing kits found online or over the counter do not evaluate sIgE antibodies; therefore, they cannot be used to diagnose an allergy.
Testing with food allergen components is primarily a type of specific IgE blood test that evaluates whether you may be allergic to a specific protein or allergen component found within a food allergen.
Think of a “whole allergen” as a completed puzzle.
Allergen components are like the puzzle pieces that make up the puzzle.
Testing with allergen components identifies which specific component of a whole allergen may be causing your reaction.
Being allergic to different types of food allergen components may cause different types of reactions. Likewise, understanding which allergen components you are allergic to can also broaden your understanding of your food allergy, such as …11, 12
Whether you may be able to tolerate baked milk or baked eggs.
How severe your peanut or tree nut allergy may be.
Whether fruit or vegetables may be cross-reacting with your pollen allergy.
Schedule an appointment with your healthcare provider today and get tested for food allergies.
Use this allergy questionnaire to help prepare you for your food allergy appointment. Once filled out, you can provide it to your clinician so that you can have an efficient and impactful conversation about your allergies