For Clinicians
Resources for Medical Professionals  
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
  Food  
  Seasonal  
  Indoor  
  Allergic Asthma  
Is your patient a candidate for specific IgE testing?
Find out 
Get detailed information on whole allergens and allergen components. 
Allergen Encyclopedia 
Ready to test a patient?
On-Demand Education 
Ragweed allergy: How to manage late-season allergies
July 2026  |  ✓  Medically Reviewed by: Gary Falcetano, PA-C, AE-C
Ragweed allergy graphic with woman sneezing
If sneezing, congestion, or itchy eyes return every year in late summer or fall, you're not alone. Ragweed pollen is a common trigger of allergy symptoms and a leading cause of seasonal allergic rhinitis in many parts of the world.1
This article explains what ragweed allergy is, why symptoms often peak in late summer and fall, how it is diagnosed and what you can do to manage your symptoms better.
What is ragweed and ragweed allergy?
Common ragweed is a weed that grows in many parts of North America, Europe, and other regions worldwide. Although there are several species, common ragweed is one of the most common triggers of seasonal allergy symptoms.2 A ragweed allergy occurs when the immune system mistakenly identifies ragweed pollen as harmful, triggering an allergic reaction.
Ragweed pollen season usually begins in late summer and continues through fall. The exact timing depends on where you live, local climate, and pollen patterns. Pollen levels often peak before declining as colder weather arrives.
Ragweed is a common trigger of seasonal allergy symptoms because it produces large amounts of pollen. A single ragweed plant can release up to 1 billion pollen grains in a season. These tiny pollen particles can travel long distances on the wind, so exposure is possible even if ragweed isn’t growing nearby.2
Symptoms and health impacts of ragweed allergy
Ragweed allergy symptoms can range from mild irritation to significant disruption of daily life. Common symptoms include:3
  • Sneezing
  • Runny nose
  • Stuffy nose
  • Itchy or watery eyes
  • Cough
These symptoms are often associated with seasonal allergic rhinitis, an allergic condition triggered by airborne allergens like ragweed pollen.
For some people, exposure to ragweed can also affect the lungs. If you have asthma, exposure during peak pollen season may worsen symptoms such as coughing, wheezing, chest tightness, or shortness of breath.4  
Ragweed allergy or a cold?
Because symptoms can overlap, it is sometimes difficult to tell whether you have a ragweed allergy or a cold.
Ragweed allergy symptoms3 Common cold symptoms3
  • Itchy eyes common
  • Sneezing common
  • Runny nose common
  • Symptoms follow pollen seasons
  • No fever
  • Symptoms may last weeks or months
  • Itchy eyes uncommon
  • Sneezing possible
  • Runny nose common
  • Symptoms can occur at any time
  • Fever may occur
  • Usually resolves within one to two weeks
How ragweed allergy is diagnosed
Understanding what's causing your symptoms is the first step toward finding the right treatment plan. Your healthcare provider will ask about your symptoms, medical history and when they occur. If ragweed allergy is suspected, allergy testing can help determine whether ragweed pollen is one of your allergy triggers.
Not sure if it's a ragweed allergy?
Complete our allergy symptom questionnaire to summarize your symptoms and help you prepare for a conversation with your healthcare provider.
Blood testing for ragweed allergy
Blood allergy testing can help identify whether ragweed pollen may be contributing to your symptoms. The test requires a simple blood sample and measures specific IgE antibodies, which are produced by the immune system in response to particular allergens.
Blood allergy testing may be particularly helpful when:
  • Additional information is needed to better understand possible allergy triggers
  • Symptoms occur during specific times of the year, such as ragweed pollen season
  • Multiple allergy triggers may be contributing to symptoms
  • A healthcare provider wants more information to help guide diagnosis and management
Results from a specific IgE blood test can help identify whether your immune system is reacting to ragweed and other potential allergens. Your healthcare provider will interpret the results alongside your symptoms, medical history, and exposure patterns to help determine whether these sensitivities may be contributing to your symptoms.
How to get an allergy test for ragweed
If you think ragweed may be causing your symptoms, talk with your healthcare provider. They can recommend the most appropriate testing based on your symptoms and medical history.
Learn more about how to get an allergy test and complete the allergy symptom questionnaire to help you prepare for a conversation with your healthcare provider.
Frequently asked questions
Ragweed pollen season typically begins in early August, peaks in September and October, and declines after the first hard frost. The exact timing depends on where you live. 
Ragweed allergy symptoms often include sneezing, itchy eyes, and a runny nose without fever. Symptoms also tend to recur during the same pollen seasons each year and may last longer than a cold. Allergy testing, combined with your symptom history, can help determine whether ragweed pollen is contributing to your reactions and support the development of an appropriate treatment plan.
Yes. If you’re allergic to ragweed, exposure to ragweed pollen can trigger or worsen asthma symptoms, especially during peak pollen season. Symptoms may include coughing, wheezing, chest tightness, or shortness of breath. If you notice that your asthma symptoms worsen during late summer or fall, talk with your healthcare provider.
Some people with ragweed allergy may develop pollen-food allergy syndrome after eating certain fruits or vegetables. This can cause itching or tingling in the mouth, lips, or throat.4 Your healthcare provider can help determine whether these reactions are related to a pollen allergy.
Although it's difficult to avoid pollen completely, these steps may help reduce your exposure during ragweed season:
  • Keep windows closed on high-pollen days
  • Check local pollen forecasts before spending time outdoors
  • Shower and change your clothes after being outside⁵
Yes. Although many allergies begin during childhood, ragweed allergy can develop at any age. If you notice allergy symptoms returning during late summer or fall, talk with your healthcare provider about whether allergy testing may be appropriate.
  1. Lake IR, Jones NR, Agnew M, et al. Climate Change and Future Pollen Allergy in Europe. Environ Health Perspect. 2017;125(3):385-391.
  2. Essl F, et al. 2015. Biological Flora of the British Isles: Ambrosia artemisiifolia. Journal of Ecology. 103:1069–1098.
  3. Wise SK, Lin SY, Toskala E, et al. International Consensus Statement on Allergy and Rhinology: Allergic Rhinitis. Int Forum Allergy Rhinol. 2023;13:293-859.
  4. Werfel T, Asero R, Ballmer-Weber BK, et al. Position paper of the EAACI: food allergy due to immunological cross-reactions with common inhalant allergens. Allergy. 2015;70:1079–1090.
  5. Environmental management of pediatric asthma. Guidelines for health care providers. National Guideline Clearinghouse. 2008