Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
September 2024 | ✓ Medically Reviewed by: Gary Falcetano, PA-C, AE-C & Magnus Borres, Director of Medical Affairs
If you’ve ever had an itchy mouth or burning lips after eating a piece of fresh fruit or some nuts, you may have experienced an allergic reaction. This is especially true if you have certain pollen allergies. How are pollen and food sensitivity connected? Proteins in both elicit a similar reaction in patients with pollen allergies.1 That tingling or burning sensation is called pollen-food allergy syndrome (PFAS).
PFAS is caused by cross-reactivity, which is when our bodies recognize proteins in one substance, such as pollen, as being similar to proteins found in another substance, like certain foods.2
For patients with an allergy to pollen proteins, their bodies react when they encounter a comparably shaped protein in food. These proteins are known as Class 2 allergens because they break down when heated or during digestion, so they may cause a reaction when they first touch someone’s mouth.3
Cross-reactivity caused by PFAS is more common than you may think — it’s estimated that over 60% of food allergies are cross-reactions between food and inhaled allergens.1
PFAS symptoms are generally mild, but in certain cases, it can result in life-threatening anaphylaxis, making it important for patients to understand their triggers.4 Allergy patients may not even think to tell their healthcare provider about these reactions to food because they seem unrelated to seasonal allergies.4
A specific IgE blood test is an important part of determining allergic triggers, so if you’ve experienced PFAS symptoms, talk to your healthcare provider about testing and an allergy-focused medical history profile.
Certain plant families have similar-shaped proteins that cause cross-reactivity, making it vital to understand if you’re sensitized to certain pollens.
Pollens from the Betulacaea family of trees include, alder, and hazel trees and are a common cause of cross-reactivity with food. Studies have found that up to 70% of patients with a birch allergy also have PFAS.2
People with these allergies often experience PFAS when eating stone fruits, or more specifically, fruits from the Rosacaea family, which includes apples, pears, peaches, plums, cherries, and apricots. Kiwis, bananas, and strawberries also cause PFAS symptoms, as do vegetables including potatoes, celery and carrots.2,4
Nuts are another important trigger of cross-reactions, particularly hazelnuts, almonds and walnuts.2 Legumes such as beans, peanuts, lentils and even wheat, can cause PFAS in patients with birch allergies.4
Alder trees are also a member of the birch family, and patients with alder pollen allergies can have similar PFAS triggers — namely, stone fruits like apples, cherries, and peaches; other fruits and vegetables like strawberries and celery; and nuts including hazelnuts, almonds, and walnuts.4
Why are there so many cross-reactions between pollens in the birch family and other foods? Because those fruits and vegetables, or more specifically, the plants they come from, have proteins similar to the allergy-inducing Bet v 1 protein in birch pollen.6 In addition to the Rosacaea family, the Apiacaea family, which includes celery and carrots, has proteins that are similar to Bet v 1.5
Sensitivity to weed pollen can cause PFAS with kiwis and bananas, and weed pollen is also noted for cross-reactivity with the Cucurbitaceae family. This plant family includes many types of melons as well as zucchini and cucumber. One study found that 50% of ragweed patients were sensitized to a member of this plant family.5
Patients with allergies to ragweed and other weed pollens may experience PFAS from honeydew and watermelons. They may also have symptoms after eating tomatoes, cucumbers, or squash. For people with allergies to grass pollen, melons and tomatoes are also PFAS triggers.4
Patients with ragweed pollen allergies may have cross-reactivity with sunflower seeds, but otherwise, nut allergies are not associated with weed pollen allergies. Grass however, show an association with peanut cross-reactivity, although peanuts are technically a legume, not a nut.6
The proteins that cause PFAS symptoms can usually be neutralized by cooking, except in the case of celery and strawberries.1 This means that consuming raw food, as fruits typically are eaten, is when PFAS symptoms are most likely to occur or are worse.
By getting a specific IgE blood test, you and your healthcare provider can build a fuller picture of your allergic triggers and develop a plan to manage them, which might include avoiding the food.
PFAS can be a minor irritation or a more serious health risk, but with allergy testing and medical history information, you can equip yourself to avoid your triggers and still enjoy fresh foods — and not the ones that will cause an itchy mouth.
Track allergy symptoms and prepare for a visit with a healthcare provider.
Learn about specific allergens, including common symptoms, management, and relief.
Are you a healthcare provider? Get comprehensive information on hundreds of whole allergens and allergen components.