Product References
Microbiota-Derived Metabolites Suppress Arthritis by Amplifying Aryl-Hydrocarbon Receptor Activation in Regulatory B Cells.
Cell metabolism
Rosser EC,Piper CJM,Matei DE,Blair PA,Rendeiro AF,Orford M,Alber DG,Krausgruber T,Catalan D,Klein N,Manson JJ,Drozdov I,Bock C,Wedderburn LR,Eaton S,Mauri C
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunohistochemistry
Tue Apr 07 00:00:00 EDT 2020
Single cell and tissue-transcriptomic analysis of murine bladders reveals age- and TNFα-dependent but microbiota-independent tertiary lymphoid tissue formation.
Mucosal immunology
Ligon MM,Wang C,DeJong EN,Schulz C,Bowdish DME,Mysorekar IU
13-5902-82 was used in Immunohistochemistry (Paraffin) to indicate that chronic, age-associated inflammation underlies a fundamental alteration to the bladder and establishes a resource for further investigation of the bladder immune system in homeostasis, aging, and disease.
Sun Nov 01 00:00:00 EDT 2020
αv Integrins regulate germinal center B cell responses through noncanonical autophagy.
The Journal of clinical investigation
Raso F,Sagadiev S,Du S,Gage E,Arkatkar T,Metzler G,Stuart LM,Orr MT,Rawlings DJ,Jackson SW,Lacy-Hulbert A,Acharya M
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Fri Aug 31 00:00:00 EDT 2018
Anti‑IL‑39 (IL‑23p19/Ebi3) polyclonal antibodies ameliorate autoimmune symptoms in lupus‑like mice.
Molecular medicine reports
Wang X,Zhang Y,Wang Z,Liu X,Zhu G,Han G,Chen G,Hou C,Wang T,Shen B,Li Y,Xiao H,Ma N,Wang R
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
B cell activation and plasma cell differentiation are inhibited by de novo DNA methylation.
Nature communications
Barwick BG,Scharer CD,Martinez RJ,Price MJ,Wein AN,Haines RR,Bally APR,Kohlmeier JE,Boss JM
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunofluorescence
Tue May 15 00:00:00 EDT 2018
Act1 is a negative regulator in T and B cells via direct inhibition of STAT3.
Nature communications
Zhang CJ,Wang C,Jiang M,Gu C,Xiao J,Chen X,Martin BN,Tang F,Yamamoto E,Xian Y,Wang H,Li F,Sartor RB,Smith H,Husni ME,Shi FD,Gao J,Carman J,Dongre A,McKarns SC,Coppieters K,Jørgensen TN,Leonard WJ,Li X
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Mon Jul 16 00:00:00 EDT 2018
Peli1 negatively regulates noncanonical NF-κB signaling to restrain systemic lupus erythematosus.
Nature communications
Liu J,Huang X,Hao S,Wang Y,Liu M,Xu J,Zhang X,Yu T,Gan S,Dai D,Luo X,Lu Q,Mao C,Zhang Y,Shen N,Li B,Huang M,Zhu X,Jin J,Cheng X,Sun SC,Xiao Y
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Mon Mar 19 00:00:00 EDT 2018
B Cell Receptor Crosslinking Augments Germinal Center B Cell Selection when T Cell Help Is Limiting.
Cell reports
Turner JS,Ke F,Grigorova IL
13-5902-82 was used in Flow cytometry/Cell sorting to investigate how T cells influence germinal center B cell expansion and plasmablast formation.
Tue Nov 06 00:00:00 EST 2018
Deficiency in Calcium-Binding Protein S100A4 Impairs the Adjuvant Action of Cholera Toxin.
Frontiers in immunology
Sun JB,Holmgren J,Larena M,Terrinoni M,Fang Y,Bresnick AR,Xiang Z
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunofluorescence
Wed Nov 20 00:00:00 EST 2019
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Artery Tertiary Lymphoid Organs Control Multilayered Territorialized Atherosclerosis B-Cell Responses in Aged ApoE-/- Mice.
Arteriosclerosis, thrombosis, and vascular biology
Srikakulapu P,Hu D,Yin C,Mohanta SK,Bontha SV,Peng L,Beer M,Weber C,McNamara CA,Grassia G,Maffia P,Manz RA,Habenicht AJ
13-5902 was used in Flow cytometry/Cell sorting to study how artery tertiary lymphoid organs can orchestrate B-cell responses in the diseased aorta.
Wed Jun 01 00:00:00 EDT 2016
Monocyte- and Neutrophil-Derived CXCL10 Impairs Efficient Control of Blood-Stage Malaria Infection and Promotes Severe Disease.
Journal of immunology (Baltimore, Md. : 1950)
Ioannidis LJ,Nie CQ,Ly A,Ryg-Cornejo V,Chiu CY,Hansen DS
13-5902 was used in Immunofluorescence to study how some inflammatory responses elicited in response to malaria infection contribute to the development of high parasite densities involved in the induction of severe disease in target organs.
Mon Feb 01 00:00:00 EST 2016
Limited clonal relatedness between gut IgA plasma cells and memory B cells after oral immunization.
Nature communications
Bemark M,Hazanov H,Strömberg A,Komban R,Holmqvist J,Köster S,Mattsson J,Sikora P,Mehr R,Lycke NY
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunofluorescence
Tue Sep 06 00:00:00 EDT 2016
Limited clonal relatedness between gut IgA plasma cells and memory B cells after oral immunization.
Nature communications
Bemark M,Hazanov H,Strömberg A,Komban R,Holmqvist J,Köster S,Mattsson J,Sikora P,Mehr R,Lycke NY
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunofluorescence
Tue Sep 06 00:00:00 EDT 2016
Diversity of the CD4 T Cell Alloresponse: The Short and the Long of It.
Cell reports
Ali JM,Negus MC,Conlon TM,Harper IG,Qureshi MS,Motallebzadeh R,Willis R,Saeb-Parsy K,Bolton EM,Bradley JA,Pettigrew GJ
13-5902 was used in Immunohistochemistry to indicate that direct and indirect-pathway CD4 T cell alloresponses, against MHC II alloantigens, are curtailed by elimination of donor hematopoietic antigen-presenting cells.
Tue Feb 09 00:00:00 EST 2016
The G protein-coupled receptor P2RY8 and follicular dendritic cells promote germinal center confinement of B cells, whereas S1PR3 can contribute to their dissemination.
The Journal of experimental medicine
Muppidi JR,Lu E,Cyster JG
13-5902 was used in Immunohistochemistry to study factors governing B cell migration to and from germinal centres.
Mon Dec 14 00:00:00 EST 2015
Interleukin (IL)-21 promotes intestinal IgA response to microbiota.
Mucosal immunology
Cao AT,Yao S,Gong B,Nurieva RI,Elson CO,Cong Y
13-5902 was used in Immunofluorescence-paraffin to investigate the role of Th17 and Tfh cytokines in regulating the mucosal IgA response to enteric microbiota.
Tue Sep 01 00:00:00 EDT 2015
DNA vaccine molecular adjuvants SP-D-BAFF and SP-D-APRIL enhance anti-gp120 immune response and increase HIV-1 neutralizing antibody titers.
Journal of virology
Gupta S,Clark ES,Termini JM,Boucher J,Kanagavelu S,LeBranche CC,Abraham S,Montefiori DC,Khan WN,Stone GW
13-5902 was used in Flow cytometry/Cell sorting to demonstrate that a DNA vaccine encoding soluble adjuvant multitrimers, IL-12p70, and membrane-bound HIV-1 Env gp140 induces HIV-1 neutralising antibodies in mice.
Wed Apr 01 00:00:00 EDT 2015
BILL-cadherin/cadherin-17 contributes to the survival of memory B cells.
PloS one
Funakoshi S,Shimizu T,Numata O,Ato M,Melchers F,Ohnishi K
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Wed Sep 16 00:00:00 EDT 2015
Establishment of murine gammaherpesvirus latency in B cells is not a stochastic event.
PLoS pathogens
Decalf J,Godinho-Silva C,Fontinha D,Marques S,Simas JP
13-5902 was used in Immunofluorescence to study whether B cell receptor specificity plays a role in B cell susceptibility to Murid γ-herpesvirus-4 latency.
Tue Jul 01 00:00:00 EDT 2014
Siglec-G deficiency leads to more severe collagen-induced arthritis and earlier onset of lupus-like symptoms in MRL/lpr mice.
Journal of immunology (Baltimore, Md. : 1950)
Bökers S,Urbat A,Daniel C,Amann K,Smith KG,Espéli M,Nitschke L
13-5902 was used in Flow cytometry/Cell sorting to study how loss of the inhibitory receptor Siglec-G led to a moderate exacerbation of disease severity and early onset in both collagen-induced arthritis and spontaneous lupus nephritis.
Tue Apr 01 00:00:00 EDT 2014
Mutation of POLB causes lupus in mice.
Cell reports
Senejani AG,Liu Y,Kidane D,Maher SE,Zeiss CJ,Park HJ,Kashgarian M,McNiff JM,Zelterman D,Bothwell AL,Sweasy JB
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Flow Cytometry
Thu Jan 16 00:00:00 EST 2014
Addendum: IgE+ memory B cells and plasma cells generated through a germinal-center pathway.
Nature immunology
Talay O,Yan D,Brightbill HD,Straney EE,Zhou M,Ladi E,Lee WP,Egen JG,Austin CD,Xu M,Wu LC
13-5902 was used in Flow cytometry/Cell sorting to identify three distinct germinal centre and memory B cell populations in IgE-GFP reporter mice.
Sun Dec 01 00:00:00 EST 2013
Natural IgG antibodies provide innate protection against ficolin-opsonized bacteria.
The EMBO journal
Panda S,Zhang J,Tan NS,Ho B,Ding JL
Published figure using GL7 monoclonal antibody (Product # 13-5902-82) in Immunofluorescence
Wed Nov 13 00:00:00 EST 2013
Re-utilization of germinal centers in multiple Peyer's patches results in highly synchronized, oligoclonal, and affinity-matured gut IgA responses.
Mucosal immunology
Bergqvist P,Stensson A,Hazanov L,Holmberg A,Mattsson J,Mehr R,Bemark M,Lycke NY
13-5902 was used in Immunohistochemistry to follow gut immunoglobulin A responses to oral hapten conjugates at the cellular and molecular level.
Tue Jan 01 00:00:00 EST 2013
Galectin 1 modulates plasma cell homeostasis and regulates the humoral immune response.
Journal of immunology (Baltimore, Md. : 1950)
Anginot A,Espeli M,Chasson L,Mancini SJ,Schiff C
13-5902 was used in Flow cytometry/Cell sorting to elucidate the pattern of GAL1 expression and the function of GAL1 in B cells.
Sat Jun 01 00:00:00 EDT 2013
Direct reduction of antigen receptor expression in polyclonal B cell populations developing in vivo results in light chain receptor editing.
Journal of immunology (Baltimore, Md. : 1950)
Shen S,Manser T
13-5902 was used in Flow cytometry/Cell sorting to test the role of BCR expression plays in receptor editing in an polyclonal, in vivo population of B cells.
Sun Jan 01 00:00:00 EST 2012
Programming the magnitude and persistence of antibody responses with innate immunity.
Nature
Kasturi SP,Skountzou I,Albrecht RA,Koutsonanos D,Hua T,Nakaya HI,Ravindran R,Stewart S,Alam M,Kwissa M,Villinger F,Murthy N,Steel J,Jacob J,Hogan RJ,García-Sastre A,Compans R,Pulendran B
13-5902 was used in Flow cytometry/Cell sorting to assess the impact of vaccination on antibody responses and to identify the critical parameters of innate immunity that program antibody responses.
Thu Feb 24 00:00:00 EST 2011
DNA methyltransferase 1 and DNA methylation patterning contribute to germinal center B-cell differentiation.
Blood
Shaknovich R,Cerchietti L,Tsikitas L,Kormaksson M,De S,Figueroa ME,Ballon G,Yang SN,Weinhold N,Reimers M,Clozel T,Luttrop K,Ekstrom TJ,Frank J,Vasanthakumar A,Godley LA,Michor F,Elemento O,Melnick A
13-5902 was used in Flow cytometry/Cell sorting to show a marked shift in DNA methylation patterning in germinal center B cells versus resting/naive B cells.
Thu Sep 29 00:00:00 EDT 2011
IL-21 is pivotal in determining age-dependent effectiveness of immune responses in a mouse model of human hepatitis B.
The Journal of clinical investigation
Publicover J,Goodsell A,Nishimura S,Vilarinho S,Wang ZE,Avanesyan L,Spolski R,Leonard WJ,Cooper S,Baron JL
13-5902 was used in Flow cytometry/Cell sorting to characterise an animal model that mimics important aspects of the age-related dichotomy in human HBV infection outcome.
Tue Mar 01 00:00:00 EST 2011
Microbiota-induced tertiary lymphoid tissues aggravate inflammatory disease in the absence of RORgamma t and LTi cells.
The Journal of experimental medicine
Lochner M,Ohnmacht C,Presley L,Bruhns P,Si-Tahar M,Sawa S,Eberl G
13-5902 was used in Immunofluorescence to investigate potential for intestinal immunodeficiency compensation by increased lymphoid tissue genesis.
Mon Jan 17 00:00:00 EST 2011
Memory B cells, but not long-lived plasma cells, possess antigen specificities for viral escape mutants.
The Journal of experimental medicine
Purtha WE,Tedder TF,Johnson S,Bhattacharya D,Diamond MS
13-5902 was used in Flow cytometry/Cell sorting to study the ability of memory B cells to target mutant viral epitopes.
Mon Dec 19 00:00:00 EST 2011
Magnetic-based purification of untouched mouse germinal center B cells for ex vivo manipulation and biochemical analysis.
Nature protocols
Cato MH,Yau IW,Rickert RC
13-5902 was used in Magnetic cell separation to provide a protocol for a novel magnetic bead separation scheme for the purification of mature B cells for biochemical assessment.
Thu Jun 09 00:00:00 EDT 2011
Identification and characterization of a lupus suppressor 129 locus on chromosome 3.
Journal of immunology (Baltimore, Md. : 1950)
Carlucci F,Fossati-Jimack L,Dumitriu IE,Heidari Y,Walport MJ,Szajna M,Baruah P,Garden OA,Cook HT,Botto M
13-5902 was used in Immunohistochemistry to identify that 129-Sle18 represents a novel, non-MHC lupus-suppressor locus, likely functionally modifying B cells, leading to lupus resistance.
Tue Jun 01 00:00:00 EDT 2010
Human B-lymphocytes express alpha2-6-sialylated 6-sulfo-N-acetyllactosamine serving as a preferred ligand for CD22/Siglec-2.
The Journal of biological chemistry
Kimura N,Ohmori K,Miyazaki K,Izawa M,Matsuzaki Y,Yasuda Y,Takematsu H,Kozutsumi Y,Moriyama A,Kannagi R
Fri Nov 02 00:00:00 EDT 2007