Product References
Alpha1-antitrypsin protects the immature mouse brain following hypoxic-ischemic injury.
Frontiers in cellular neuroscience
Zhang S,Li W,Xu Y,Li T,Ek J,Zhang X,Wang Y,Song J,Zhu C,Wang X
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Mar 24 00:00:00 EDT 2023
Endocytic membrane repair by ESCRT-III controls antigen export to the cytosol during antigen cross-presentation.
Cell reports
Gros M,Segura E,Rookhuizen DC,Baudon B,Heurtebise-Chrétien S,Burgdorf N,Maurin M,Kapp EA,Simpson RJ,Kozik P,Villadangos JA,Bertrand MJM,Burbage M,Amigorena S
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunocytochemistry
Tue Aug 16 00:00:00 EDT 2022
Amyloid-beta and tau pathologies act synergistically to induce novel disease stage-specific microglia subtypes.
Molecular neurodegeneration
Kim DW,Tu KJ,Wei A,Lau AJ,Gonzalez-Gil A,Cao T,Braunstein K,Ling JP,Troncoso JC,Wong PC,Blackshaw S,Schnaar RL,Li T
14-5301-82 was used in Immunohistochemistry to suggest that both Aβ and tau pathologies are required for the disease stage-specific induction of early-stage AD-Associated Microglia (EADAM) and late-stage AD-Associated Microglia (LADAM), and reveal Siglecs as biomarkers of Alzheimer's disease progression.
Sat Dec 17 00:00:00 EST 2022
Phospholipid nanoparticles: Therapeutic potentials against atherosclerosis via reducing cholesterol crystals and inhibiting inflammation.
EBioMedicine
Luo Y,Guo Y,Wang H,Yu M,Hong K,Li D,Li R,Wen B,Hu D,Chang L,Zhang J,Yang B,Sun D,Schwendeman AS,Eugene Chen Y
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Wed Dec 01 00:00:00 EST 2021
KLF11 protects against abdominal aortic aneurysm through inhibition of endothelial cell dysfunction.
JCI insight
Zhao G,Chang Z,Zhao Y,Guo Y,Lu H,Liang W,Rom O,Wang H,Sun J,Zhu T,Fan Y,Chang L,Yang B,Garcia-Barrio MT,Chen YE,Zhang J
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
KLF11 protects against abdominal aortic aneurysm through inhibition of endothelial cell dysfunction.
JCI insight
Zhao G,Chang Z,Zhao Y,Guo Y,Lu H,Liang W,Rom O,Wang H,Sun J,Zhu T,Fan Y,Chang L,Yang B,Garcia-Barrio MT,Chen YE,Zhang J
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunocytochemistry
Mon Mar 08 00:00:00 EST 2021
Quantitative characterization of extracellular vesicle uptake and content delivery within mammalian cells.
Nature communications
Bonsergent E,Grisard E,Buchrieser J,Schwartz O,Théry C,Lavieu G
14-5301-82 was used in Immunocytochemistry to show that extracellular vesicle content release requires endosomal acidification with the potential involvement of membrane fusion.
Thu Mar 25 00:00:00 EDT 2021
Overexpression of Galectin 3 in Pancreatic β Cells Amplifies β-Cell Apoptosis and Islet Inflammation in Type-2 Diabetes in Mice.
Frontiers in endocrinology
Petrovic I,Pejnovic N,Ljujic B,Pavlovic S,Miletic Kovacevic M,Jeftic I,Djukic A,Draginic N,Andjic M,Arsenijevic N,Lukic ML,Jovicic N
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Wed Feb 03 00:00:00 EST 2021
Inhibition of p38mapk Reduces Adipose Tissue Inflammation in Aging Mediated by Arginase-II.
Pharmacology
Huang J,Liu C,Ming XF,Yang Z
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Wed Feb 24 00:00:00 EST 2021
Smooth muscle-derived progenitor cell myofibroblast differentiation through KLF4 downregulation promotes arterial remodeling and fibrosis.
JCI insight
Lu S,Jolly AJ,Strand KA,Dubner AM,Mutryn MF,Moulton KS,Nemenoff RA,Majesky MW,Weiser-Evans MC
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Thu Dec 03 00:00:00 EST 2020
Smooth muscle-derived progenitor cell myofibroblast differentiation through KLF4 downregulation promotes arterial remodeling and fibrosis.
JCI insight
Lu S,Jolly AJ,Strand KA,Dubner AM,Mutryn MF,Moulton KS,Nemenoff RA,Majesky MW,Weiser-Evans MC
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Thu Dec 03 00:00:00 EST 2020
Macrophage NCOR1 Deficiency Ameliorates Myocardial Infarction and Neointimal Hyperplasia in Mice.
Journal of the American Heart Association
Du LJ,Sun JY,Zhang WC,Wang YL,Zhu H,Liu T,Gao MZ,Zheng C,Zhang YY,Liu Y,Liu Y,Shao S,Zhang XQ,Leng Q,Auwerx J,Duan SZ
14-5301-85 was used in Immunohistochemistry (Paraffin) to demonstrate that NCOR1 is a critical regulator of macrophage inflammation and proliferation and that deficiency of NCOR1 in macrophages attenuates MI and neointimal hyperplasia.
Tue Aug 04 00:00:00 EDT 2020
Macrophage NCOR1 Deficiency Ameliorates Myocardial Infarction and Neointimal Hyperplasia in Mice.
Journal of the American Heart Association
Du LJ,Sun JY,Zhang WC,Wang YL,Zhu H,Liu T,Gao MZ,Zheng C,Zhang YY,Liu Y,Liu Y,Shao S,Zhang XQ,Leng Q,Auwerx J,Duan SZ
14-5301-85 was used in Immunohistochemistry (Paraffin) to demonstrate that NCOR1 is a critical regulator of macrophage inflammation and proliferation and that deficiency of NCOR1 in macrophages attenuates MI and neointimal hyperplasia.
Tue Aug 04 00:00:00 EDT 2020
Foreign body responses in mouse central nervous system mimic natural wound responses and alter biomaterial functions.
Nature communications
O¿Shea TM,Wollenberg AL,Kim JH,Ao Y,Deming TJ,Sofroniew MV
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Dec 04 00:00:00 EST 2020
Foreign body responses in mouse central nervous system mimic natural wound responses and alter biomaterial functions.
Nature communications
O¿Shea TM,Wollenberg AL,Kim JH,Ao Y,Deming TJ,Sofroniew MV
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Dec 04 00:00:00 EST 2020
Ultrastructural alteration of pulmonary tissue under conditions of high oxygen concentration.
International journal of clinical and experimental pathology
Yano H,Kuroda A,Okada H,Tomita H,Suzuki K,Takada C,Fukuda H,Kawasaki Y,Muraki I,Wakayama Y,Kano S,Tamaoki Y,Nishio A,Inagawa R,Sampei S,Kamidani R,Kakino Y,Yasuda R,Kitagawa Y,Fukuta T,Miyake T,Kanda N,Miyazaki N,Doi T,Yoshida T,Suzuki A,Yoshida S,Ogura S
14-5301 was used in Immunohistochemistry to document the ultrastructural morphology of hyperoxic acute lung injury including endothelial glycocalyx degradation.
Tue Jan 12 00:00:00 EST 2021
An Orally Active Galectin-3 Antagonist Inhibits Lung Adenocarcinoma Growth and Augments Response to PD-L1 Blockade.
Cancer research
Vuong L,Kouverianou E,Rooney CM,McHugh BJ,Howie SEM,Gregory CD,Forbes SJ,Henderson NC,Zetterberg FR,Nilsson UJ,Leffler H,Ford P,Pedersen A,Gravelle L,Tantawi S,Schambye H,Sethi T,MacKinnon AC
14-5301-82 was used in Western Blot to explore the role of β-galactoside-binding lectin galectin-3 in patients with non-small cell lung cancer.
Mon Apr 01 00:00:00 EDT 2019
Xanthohumol and 8-prenylnaringenin reduce type 2 diabetes-associated oxidative stress by downregulating galectin-3.
Porto biomedical journal
Luís C,Costa R,Rodrigues I,Castela Â,Coelho P,Guerreiro S,Gomes J,Reis C,Soares R
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Sun Apr 10 00:00:00 EDT 2022
Xanthohumol and 8-prenylnaringenin reduce type 2 diabetes-associated oxidative stress by downregulating galectin-3.
Porto biomedical journal
Luís C,Costa R,Rodrigues I,Castela Â,Coelho P,Guerreiro S,Gomes J,Reis C,Soares R
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Sun Apr 10 00:00:00 EDT 2022
Time-course analysis of cardiac and serum galectin-3 in viral myocarditis after an encephalomyocarditis virus inoculation.
PloS one
Noguchi K,Tomita H,Kanayama T,Niwa A,Hatano Y,Hoshi M,Sugie S,Okada H,Niwa M,Hara A
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Nov 01 00:00:00 EDT 2019
Time-course analysis of cardiac and serum galectin-3 in viral myocarditis after an encephalomyocarditis virus inoculation.
PloS one
Noguchi K,Tomita H,Kanayama T,Niwa A,Hatano Y,Hoshi M,Sugie S,Okada H,Niwa M,Hara A
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Nov 01 00:00:00 EDT 2019
Myeloid-specific deletion of Zfp36 protects against insulin resistance and fatty liver in diet-induced obese mice.
American journal of physiology. Endocrinology and metabolism
Caracciolo V,Young J,Gonzales D,Ni Y,Flowers SJ,Summer R,Waldman SA,Kim JK,Jung DY,Noh HL,Kim T,Blackshear PJ,O'Connell D,Bauer RC,Kallen CB
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Western Blot
Mon Oct 01 00:00:00 EDT 2018
Myeloid-specific deletion of Zfp36 protects against insulin resistance and fatty liver in diet-induced obese mice.
American journal of physiology. Endocrinology and metabolism
Caracciolo V,Young J,Gonzales D,Ni Y,Flowers SJ,Summer R,Waldman SA,Kim JK,Jung DY,Noh HL,Kim T,Blackshear PJ,O'Connell D,Bauer RC,Kallen CB
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Western Blot
Mon Oct 01 00:00:00 EDT 2018
T-Cell Mineralocorticoid Receptor Controls Blood Pressure by Regulating Interferon-Gamma.
Circulation research
Sun XN,Li C,Liu Y,Du LJ,Zeng MR,Zheng XJ,Zhang WC,Liu Y,Zhu M,Kong D,Zhou L,Lu L,Shen ZX,Yi Y,Du L,Qin M,Liu X,Hua Z,Sun S,Yin H,Zhou B,Yu Y,Zhang Z,Duan SZ
14-5301 was used in Immunohistochemistry to determine the role of T-cell mineralocorticoid receptor in blood pressure regulation and explore the mechanism.
Fri May 12 00:00:00 EDT 2017
T-Cell Mineralocorticoid Receptor Controls Blood Pressure by Regulating Interferon-Gamma.
Circulation research
Sun XN,Li C,Liu Y,Du LJ,Zeng MR,Zheng XJ,Zhang WC,Liu Y,Zhu M,Kong D,Zhou L,Lu L,Shen ZX,Yi Y,Du L,Qin M,Liu X,Hua Z,Sun S,Yin H,Zhou B,Yu Y,Zhang Z,Duan SZ
14-5301 was used in Immunohistochemistry to determine the role of T-cell mineralocorticoid receptor in blood pressure regulation and explore the mechanism.
Fri May 12 00:00:00 EDT 2017
Macrophage Proliferation Sustains Adipose Tissue Inflammation in Formerly Obese Mice.
Diabetes
Zamarron BF,Mergian TA,Cho KW,Martinez-Santibanez G,Luan D,Singer K,DelProposto JL,Geletka LM,Muir LA,Lumeng CN
14-5301 was used in Immunohistochemistry-immunofluorescence to study the mechanisms by which meta-inflammation resolves during weight loss, by examining adipose tissue leukocytes in mice after withdrawal of a high-fat diet.
Wed Feb 01 00:00:00 EST 2017
SCHLAFEN 5 expression correlates with intestinal metaplasia that progresses to gastric cancer.
Journal of gastroenterology
Companioni Nápoles O,Tsao AC,Sanz-Anquela JM,Sala N,Bonet C,Pardo ML,Ding L,Simo O,Saqui-Salces M,Blanco VP,Gonzalez CA,Merchant JL
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Sun Jan 01 00:00:00 EST 2017
Deoxycorticosterone/Salt-Mediated Cardiac Inflammation and Fibrosis Are Dependent on Functional CLOCK Signaling in Male Mice.
Endocrinology
Fletcher EK,Morgan J,Kennaway DR,Bienvenu LA,Rickard AJ,Delbridge LMD,Fuller PJ,Clyne CD,Young MJ
14-5301 was used in Immunohistochemistry to demonstrate that the full development of MR-mediated cardiac inflammation and fibrosis is dependent on intact signaling by the circadian protein CLOCK.
Fri Sep 01 00:00:00 EDT 2017
Inhibition of sphingosine 1-phosphate signaling ameliorates murine nonalcoholic steatohepatitis.
American journal of physiology. Gastrointestinal and liver physiology
Mauer AS,Hirsova P,Maiers JL,Shah VH,Malhi H
Wed Mar 01 00:00:00 EST 2017
Mixed-lineage kinase 3 pharmacological inhibition attenuates murine nonalcoholic steatohepatitis.
JCI insight
Tomita K,Kohli R,MacLaurin BL,Hirsova P,Guo Q,Sanchez LHG,Gelbard HA,Blaxall BC,Ibrahim SH
Thu Aug 03 00:00:00 EDT 2017
Mixed-lineage kinase 3 pharmacological inhibition attenuates murine nonalcoholic steatohepatitis.
JCI insight
Tomita K,Kohli R,MacLaurin BL,Hirsova P,Guo Q,Sanchez LHG,Gelbard HA,Blaxall BC,Ibrahim SH
Thu Aug 03 00:00:00 EDT 2017
The kielin/chordin-like protein (KCP) attenuates high-fat diet-induced obesity and metabolic syndrome in mice.
The Journal of biological chemistry
Soofi A,Wolf KI,Emont MP,Qi N,Martinez-Santibanez G,Grimley E,Ostwani W,Dressler GR
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Jun 02 00:00:00 EDT 2017
The kielin/chordin-like protein (KCP) attenuates high-fat diet-induced obesity and metabolic syndrome in mice.
The Journal of biological chemistry
Soofi A,Wolf KI,Emont MP,Qi N,Martinez-Santibanez G,Grimley E,Ostwani W,Dressler GR
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Fri Jun 02 00:00:00 EDT 2017
Neuroprotection by selective neuronal deletion of Atg7 in neonatal brain injury.
Autophagy
Xie C,Ginet V,Sun Y,Koike M,Zhou K,Li T,Li H,Li Q,Wang X,Uchiyama Y,Truttmann AC,Kroemer G,Puyal J,Blomgren K,Zhu C
14-5301 was used in Immunohistochemistry to reveal that selective neuronal deletion of Atg7 is strongly protective against neuronal death and overall brain injury occurring after HI.
Tue Dec 13 00:00:00 EST 2016
Galectin-3 expression in hippocampal CA2 following transient forebrain ischemia and its inhibition by hypothermia or antiapoptotic agents.
Neuroreport
Hisamatsu K,Niwa M,Kobayashi K,Miyazaki T,Hirata A,Hatano Y,Tomita H,Hara A
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunohistochemistry
Wed Mar 23 00:00:00 EDT 2016
Cell surface galectin-3 defines a subset of chemoresistant gastrointestinal tumor-initiating cancer cells with heightened stem cell characteristics.
Cell death & disease
Ilmer M,Mazurek N,Byrd JC,Ramirez K,Hafley M,Alt E,Vykoukal J,Bresalier RS
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Flow Cytometry
Thu Aug 11 00:00:00 EDT 2016
Cell surface galectin-3 defines a subset of chemoresistant gastrointestinal tumor-initiating cancer cells with heightened stem cell characteristics.
Cell death & disease
Ilmer M,Mazurek N,Byrd JC,Ramirez K,Hafley M,Alt E,Vykoukal J,Bresalier RS
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Flow Cytometry
Thu Aug 11 00:00:00 EDT 2016
Rhadinovirus host entry by co-operative infection.
PLoS pathogens
Lawler C,Milho R,May JS,Stevenson PG
14-5301 was used in Immunofluorescence to determine how a rhadinovirus enters the lungs.
Sun Mar 01 00:00:00 EST 2015
Maternal immune activation evoked by polyinosinic:polycytidylic acid does not evoke microglial cell activation in the embryo.
Frontiers in cellular neuroscience
Smolders S,Smolders SM,Swinnen N,Gärtner A,Rigo JM,Legendre P,Brône B
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Flow Cytometry
Mon Aug 24 00:00:00 EDT 2015
Maternal immune activation evoked by polyinosinic:polycytidylic acid does not evoke microglial cell activation in the embryo.
Frontiers in cellular neuroscience
Smolders S,Smolders SM,Swinnen N,Gärtner A,Rigo JM,Legendre P,Brône B
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Flow Cytometry
Mon Aug 24 00:00:00 EDT 2015
18-HEPE, an n-3 fatty acid metabolite released by macrophages, prevents pressure overload-induced maladaptive cardiac remodeling.
The Journal of experimental medicine
Endo J,Sano M,Isobe Y,Fukuda K,Kang JX,Arai H,Arita M
14-5301 was used in Immunohistochemistry to study the ability of an eicosapentaenoic acid metabolite to inhibit macrophage-mediated proinflammatory activation of cardiac fibroblasts.
Mon Jul 28 00:00:00 EDT 2014
Imaging white adipose tissue with confocal microscopy.
Methods in enzymology
Martinez-Santibañez G,Cho KW,Lumeng CN
Published figure using Galectin 3 monoclonal antibody (Product # 14-5301-82) in Immunofluorescence
Fri Sep 19 00:00:00 EDT 2014
Molecular imaging of the paracrine proangiogenic effects of progenitor cell therapy in limb ischemia.
Circulation
Ryu JC,Davidson BP,Xie A,Qi Y,Zha D,Belcik JT,Caplan ES,Woda JM,Hedrick CC,Hanna RN,Lehman N,Zhao Y,Ting A,Lindner JR
14-5301 was used in Immunohistochemistry to test the hypothesis that treatment of limb ischemia with MAPCs promotes recovery of blood flow through the recruitment of proangiogenic monocytes.
Tue Feb 12 00:00:00 EST 2013
PD-L1 has distinct functions in hematopoietic and nonhematopoietic cells in regulating T cell responses during chronic infection in mice.
The Journal of clinical investigation
Mueller SN,Vanguri VK,Ha SJ,West EE,Keir ME,Glickman JN,Sharpe AH,Ahmed R
14-5301 was used in Immunohistochemistry on paraffin embedded tissues to demonstrate that there are distinct roles for PD-L1 on hematopoietic and nonhematopoietic cells in regulating CD8+ T cell responses.
Thu Jul 01 00:00:00 EDT 2010
Glycation of a food allergen by the Maillard reaction enhances its T-cell immunogenicity: role of macrophage scavenger receptor class A type I and II.
The Journal of allergy and clinical immunology
Ilchmann A,Burgdorf S,Scheurer S,Waibler Z,Nagai R,Wellner A,Yamamoto Y,Yamamoto H,Henle T,Kurts C,Kalinke U,Vieths S,Toda M
14-5301 was used in Flow cytometry/Cell sorting to define the T-cell immunogenicity of food advanced glycation end products, using ovalbumin as a model allergen.
Fri Jan 01 00:00:00 EST 2010
Renal vascular inflammation induced by Western diet in ApoE-null mice quantified by (19)F NMR of VCAM-1 targeted nanobeacons.
Nanomedicine : nanotechnology, biology, and medicine
Southworth R,Kaneda M,Chen J,Zhang L,Zhang H,Yang X,Razavi R,Lanza G,Wickline SA
14-5301 was used in Immunohistochemistry to design multi functional nano-particulate reporter bioprobes capable of targeting vascular cell adhesion molecule 1.
Tue Sep 01 00:00:00 EDT 2009
Identification of markers that distinguish monocyte-derived fibrocytes from monocytes, macrophages, and fibroblasts.
PloS one
Pilling D,Fan T,Huang D,Kaul B,Gomer RH
14-5301 was used in Immunohistochemistry to identify markers that can be used to discriminate between human peripheral blood monocytes, tissue macrophages, fibrocytes, and fibroblasts.
Fri Oct 16 00:00:00 EDT 2009