Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Immunocytochemistry (ICC/IF) |
0.5-2 µg/mL | View 1 publication 1 publication |
Flow Cytometry (Flow) |
0.5-2 µg/mL | - |
ELISA (ELISA) |
0.1-3 µg/mL | - |
Immunoprecipitation (IP) |
0.2-2 µg/mL | - |
Dot blot (DB) |
0.5-2 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Chemical |
Published species |
Chemical |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Clone |
RM231 |
Immunogen |
BSA-conjugated 5-methylcytosine |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.2-7.4, with 1% BSA, 50% glycerol |
Contains |
0.09% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2665309 |
This antibody reacts to 5-methylcytosine in both single-stranded and double-stranded DNA. No cross reactivity with non-methylated cytosine and hydroxymethylcytosine in DNA.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Methylation of DNA cytosine bases plays important roles in the regulation of gene transcription. It has been estimated that 60 to 90% of the cytosines in CpG dinucleotides are methylated. An increase in methyl-CpG correlates with transcriptional silencing for the whole chromosome especially in developmentally regulated genes. Methylation is believed to lead to transcriptional silencing by multiple mechanisms including alteration of transcription factor binding to promoters and alteration in chromatin structure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-mC; 5-meC; 5-Methyl-cytosine; 5mc; C5H7NO3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support