Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, Bovine and Orangutan
Kinesins, such as KIF2, are microtubule-associated motor proteins. For background information on kinesins.
Protein Aliases: hK2; kinesin heavy chain member 2A; Kinesin, heavy chain, 2; Kinesin-2; Kinesin-like protein KIF2A
Gene Aliases: KIF2; KIF2A; KNS2