Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
Protein Aliases: DDB1- and CUL4-associated factor 8; WD repeat domain 42A; WD repeat-containing protein 42A
Gene Aliases: DCAF8; H326; WDR42A