Based on 100% sequence identity, this antibody is predicted to react with Orangutan
TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.
Protein Aliases: FLJ38482; Transmembrane protein 192
Gene Aliases: TMEM192