This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
Protein Aliases: Jade family PHD finger protein 3; PHD finger protein 16; Protein Jade-3
Gene Aliases: JADE3; KIAA0215; PHF16