Formation of pseudouridine at position 39 in the anticodon stem and loop of transfer RNAs.
Protein Aliases: tRNA pseudouridine synthase 3; tRNA pseudouridine(38/39) synthase; tRNA pseudouridylate synthase 3; tRNA-uridine isomerase 3
Gene Aliases: FKSG32; PUS3