Based on 100% sequence identity, this antibody is predicted to react with Rat and Zebrafish
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
Protein Aliases: E2 ubiquitin-conjugating enzyme Z; Uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2Z; Ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; Ubiquitin-protein ligase Z; Use1
Gene Aliases: D11Moh35; HOYS7; UBE2Z