Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Rabbit
The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Protein Aliases: CFA; chaperonin cofactor A; TCP1-chaperonin cofactor A; Tubulin-folding cofactor A; Tubulin-specific chaperone A
Gene Aliases: TBCA