Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Sequence of this protein is as follows: RLDPFFKQQA VCGNAKLEAG EECDCGTEQD CALIGETCCD IATCRFKAGS NCAEGPCCEN CLFMSKERMC RPSFEECDLP EYCNGSSASC PENHYVQTGH
ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; ADAM; ADAMs; cancer/testis antigen 15; disintegrin; fertilin subunit beta; metalloendopeptidases; unnamed protein product
Gene Aliases: CRYN1; CRYN2; CT15; FTNB; PH-30b; PH30; PH30-beta
UniProt ID: (Human) Q99965
Entrez Gene ID: (Human) 2515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support