Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | View 3 publications 3 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:20-1:200 | - |
Immunocytochemistry (ICC/IF) |
- | View 1 publication 1 publication |
Immunoprecipitation (IP) |
0.5-4.0 µg | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Published species |
Human, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
AHR Fusion Protein Ag12193 (1-387 aa encoded by BC070080) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Storage conditions |
-20°C |
Immunogen sequence: MNSSSANIT YASRKRRKPV QKTVKPIPAE GIKSNPSKRH RDRLNTELDR LASLLPFPQD VINKLDKLSV LRLSVSYLRA KSFFDVALKS SPTERNGGQD NCRAANFREG LNLQEGEFLL QALNGFVLVV TTDALVFYAS STIQDYLGFQ QSDVIHQSVY ELIHTEDRAE FQRQLHWALN PSQCTESGQG IEEATGLPQT VVCYNPDQIP PENSPLMERC FICRLRCLLD NSSGFLAMNF QGKLKYLHGQ KKKGKDGSIL PPQLALFAIA TPLQPPSILE IRTKNFIFRT KHKLDFTPIG CDAKGRIVLG YTEAELCTRG SGYQFIHAAD MLYCAESHIR MIKTGESGMI VFRLLTKNNR WTWVQSNARL LYKNGRPDYI IVTQRPLT (1-387 aa encoded by BC070080)
AHR (Ah Receptor) belongs to a family of proteins comprised of its dimerization partner ARNT (HIF-1 Beta) and the Drosophila proteins PER and SIM. AHR contains an N-terminal sequence of approximately 200 amino acids termed the PAS domain. AHR, found in a variety of tissues, binds to a specific DNA enhancer sequence and initiates transcription of the mRNA for the cytochrome P-450 (CYPIA1) gene. The gene for AHR encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. AHR has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450, and its ligands included a variety of aromatic hydrocarbons. AHR is a ligand-activated helix/loop/helix transcription factor found in a variety of vertebrate species. The known ligands for AHR are foreign planar aromatic compounds, such as polycyclic aromatic compounds and halogenated aromatic compounds such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). Unlike the steroid/thyroid hormone receptors, there is no known physiological ligand for AHR. Studies indicate that in non-ligand activated cells, AHR is found complexed with HSP90 predominantly in the cytoplasm. Upon binding to an agonist, the ligand-activated AhR is believed to transform to a nuclear, DNA binding form, and this transformation process appears to involve dissociation of HSP90 from AhR followed by formation of a heterodimer with AhR nuclear translocator protein (Arnt). Diseases associated with AHR include eosinophilic fasciitis and seborrheic dermatitis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ah receptor; AH-receptor; aromatic hydrocarbon receptor; Aryl hydrocarbon receptor; bHLHe76; Class E basic helix-loop-helix protein 76; dioxin receptor
Gene Aliases: AHR; BHLHE76
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support